Skip to Content
Merck
All Photos(5)

Key Documents

156159

Sigma-Aldrich

Sodium cyanoborohydride

reagent grade, 95%

Synonym(s):

Sodium cyanotrihydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH3CN
CAS Number:
Molecular Weight:
62.84
Beilstein:
4152656
EC Number:
MDL number:
UNSPSC Code:
12352306
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Assay

95%

form

powder

reaction suitability

reagent type: reductant

mp

>242 °C (dec.) (lit.)

SMILES string

[Na+].[BH3-]C#N

InChI

1S/CH3BN.Na/c2-1-3;/h2H3;/q-1;+1

InChI key

CVDUGUOQTVTBJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium cyanoborohydride, also known as sodium cyanotrihydridoborate, is a mild reducing agent that is stable in acidic conditions. It is widely used in the selective reduction of aryl aldehydes and ketones.

Application

Reagent for selective reductions.
Sodium cyanoborohydride is used as a reagent: 
  • To convert the amino-functional Si3N4 surface to a reactive surface with aldehyde groups.
  • For coupling amino-modified oligonucleotide probes.
  • In the synthesis of a novel phenolate-bridged dilanthanum(III) complex of interest as a model for metalloproteins.
  • In the reductive amination of aldehydes and ketones, and in the reductive alkylation of amines.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 8, 3-10 (1975)
Synthetic Communications, 24, 701-701 (1994)
Tetrahedron Letters, 35, 2775-2775 (1994)
Reductive deoxygenation of aryl aldehydes and ketones and benzylic, allylic, and tertiary alcohols by zinc iodide-sodium cyanoborohydride
CK L, et al.
The Journal of Organic Chemistry, 51, 3038-3043 (1986)
The Journal of Organic Chemistry, 55, 2552-2552 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service