Skip to Content
Merck
All Photos(1)

Key Documents

PP0201

Sigma-Aldrich

Glycoprofile II, Enzymatic In-Solution N-Deglycosylation Kit

sufficient for 20 samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26
Pricing and availability is not currently available.

Quality Level

storage temp.

2-8°C

General description

Contains sufficient reagents for a minimum of 20 reactions when the sample size is between one to two mg of a typical glycoprotein.

Application

GlycoProfile II Enzymatic In-Solution N-Deglycosylation Kit was used to study the effects of glycosylation of the N-acetyl-D-galactosamine (GalNAc)-specific lectin found in Schizophyllum commune.[1]
GlycoProfile II has been optimized to provide a convenient and reproducible method to remove N-linked glycans from glycoproteins and is compatible with subsequent MALDI-TOF mass spectrometric analysis without interference from any of the reaction components.
Glycoprofile II, Enzymatic In-Solution N-Deglycosylation Kit has been used in the deglycosylation of Schizophyllum commune lectin,[2] Arabidopsis mannanase,[3] human umbilical vein endothelial cells (HUVEC) protein lysate[4] and organic solute transporter protein, Ostβ.[5]

Legal Information

GlycoProfile is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • 10× Reaction Buffer 1 mL/vial

Kit Components Also Available Separately

Product No.
Description
SDS

  • P7367PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE) 50 USDS

  • R7884Ribonuclease B from bovine pancreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE) .5 mgSDS

  • O9882Octyl β-D-glucopyranoside, BioXtra, ≥98% (GC) 100 mgSDS

  • M31482-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration) .9 mLSDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Liver,Heart

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic characterization of a glycoside hydrolase family 5 subfamily 7 (GH5\_7) mannanase from Arabidopsis thaliana
Wang Y, et al.
Planta, 239(3), 653-665 (2014)
Christen Y L Yuen et al.
Plant & cell physiology, 58(6), 1103-1117 (2017-04-27)
Members of the protein disulfide isomerase (PDI)-C subfamily are chimeric proteins containing the thioredoxin (Trx) domain of PDIs, and the conserved N- and C-terminal Pfam domains of Erv41p/Erv46p-type cargo receptors. They are unique to plants and chromalveolates. The Arabidopsis genome
Podjana Chumkhunthod et al.
Biochimica et biophysica acta, 1760(3), 326-332 (2006-03-02)
An N-acetyl-D-galactosamine (GalNAc)-specific lectin was purified from the edible mushroom, Schizophyllum commune, using affinity chromatography on a porcine stomach mucin (PSM)-Sepharose 4B column. Under reducing and non-reducing conditions, SDS-polyacrylamide gel electrophoresis gave a major band of 31.5 kDa. The Schizophyllum
Evaluation of soluble junctional adhesion molecule-A as a biomarker of human brain endothelial barrier breakdown
Haarmann A, et al.
PLoS ONE, 5(10), e13568-e13568 (2010)
Elena Schwagerus et al.
Pflugers Archiv : European journal of physiology, 467(11), 2257-2273 (2015-02-14)
Using human airway epithelial cell lines (i.e. NCI-H441 and Calu-3) as well as human alveolar epithelial type I-like (ATI) cells in primary culture, we studied the contribution of the epithelial sodium channel δ-subunit (δ-ENaC) to transepithelial sodium transport in human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service