Skip to Content
Merck
All Photos(1)

Key Documents

G2911

Sigma-Aldrich

GF 109203X

synthetic, ≥90% (HPLC)

Synonym(s):

3-(N-[Dimethylamino]propyl-3-indolyl)-4-(3-indolyl)maleimide, 3-[1-[3-(Dimethylamino)propyl]1H-indol-3-yl]-4-(1Hindol-3-yl)1H-pyrrole-2,5dione, Bisindolylmaleimide I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24N4O2
CAS Number:
Molecular Weight:
412.48
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥90% (HPLC)

solubility

DMSO: soluble (~1 mg/ml)

storage temp.

2-8°C

SMILES string

CN(C)CCCn1cc(C2=C(C(=O)NC2=O)c3c[nH]c4ccccc34)c5ccccc15

InChI

1S/C25H24N4O2/c1-28(2)12-7-13-29-15-19(17-9-4-6-11-21(17)29)23-22(24(30)27-25(23)31)18-14-26-20-10-5-3-8-16(18)20/h3-6,8-11,14-15,26H,7,12-13H2,1-2H3,(H,27,30,31)

InChI key

QMGUOJYZJKLOLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

GF 109203X has been used as a protein kinase C inhibitor in HUVECs (human umbilical vein endothelial cells), angiotensinogen gene knockout (AGT KO) mice, and in granulosa cell cultures of female rats.

Biochem/physiol Actions

A potent and selective competitive inhibitor of protein kinase C (PKC) and of glycogen synthase kinase-3 (GSK-3). Inhibits parathyroid hormone-induced Ca2+ resorption from isolated bone tissue, Staurosporine, another protein kinase inhibitor, actually enhanced Ca2+ resorption elicited by a number of agents, but GF109203X counteracted that enhancement.
A potent and selective competitive inhibitor of protein kinase C (PKC) and of glycogen synthase kinase-3 (GSK-3). In certain cells, inhibition of PKC leads to an increase in autophagy. For PKC inhibition, typically used at a concentration of 0.1-10 μM.

Quantity

For PKC inhibition, typically used at a concentration of 0.1-10 μM.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Follicle-stimulating hormone (FSH) stimulates phosphorylation and activation of protein kinase B (PKB/Akt) and serum and glucocorticoid-induced kinase (Sgk): evidence for A kinase-independent signaling by FSH in granulosa cells.
Gonzalez-Robayna IJ, et al.
Molecular Endocrinology, 14(8), 1283-1300 (2000)
Src is required for mechanical stretch-induced cardiomyocyte hypertrophy through angiotensin II type 1 receptor-dependent beta-arrestin2 pathways.
Wang S, et al.
PloS one, 9(4), e92926-e92926 (2014)
Yun Zhu et al.
Environmental toxicology and pharmacology, 48, 197-202 (2016-11-07)
Studies have shown that corticotrophin-releasing hormone (CRH) and relaxin are associated with early delivery. Our lab previously has shown the mycotoxin zeranol increases placental CRH expression. The mycotoxin is used in the farming industry to promote cattle growth, and some
Kristin Klarström Engström et al.
Scientific reports, 9(1), 2296-2296 (2019-02-21)
To prevent the onset of urosepsis and reduce mortality, a better understanding of how uropathogenic Escherichia coli (UPEC) manages to infiltrate the bloodstream through the kidneys is needed. The present study elucidates if human renal interstitial fibroblasts are part of
H2O2 Treatment of HUVECs Facilitates PKC Mediated Thr495 Phosphorylation on eNOS when Pre-treated with High Glucose Levels.
Guterbaum TJ, et al.
Journal of Metabolic Syndrome, 4(189), 2167-0943 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service