Skip to Content
Merck
All Photos(1)

Key Documents

F6127

Sigma-Aldrich

Fludrocortisone acetate

≥98%

Synonym(s):

9α-Fluoro-11β,17α,21-trihydroxy-4-pregnene-3,20-dione acetate, 9-Fluorocortisol acetate, 9-Fluorohydrocortisone acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31FO6
CAS Number:
Molecular Weight:
422.49
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

sterility

non-sterile

Assay

≥98%

form

powder

solubility

acetone: 50 mg/mL, clear, colorless to light yellow

shipped in

ambient

storage temp.

room temp

SMILES string

CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

InChI

1S/C23H31FO6/c1-13(25)30-12-19(28)22(29)9-7-16-17-5-4-14-10-15(26)6-8-20(14,2)23(17,24)18(27)11-21(16,22)3/h10,16-18,27,29H,4-9,11-12H2,1-3H3/t16-,17-,18-,20-,21-,22-,23-/m0/s1

InChI key

SYWHXTATXSMDSB-GSLJADNHSA-N

Gene Information

human ... NR3C2(4306)

Looking for similar products? Visit Product Comparison Guide

General description

Fludrocortisone acetate is secreted by adrenal cortex. It increases the reabsorption of sodium and secretion of potassium and hydrogen. Fludrocortisone is used to treat cerebral salt wasting syndrome and adrenal insufficiency. Fludrocortisone improves the sensitivity of blood vessels to catecholamines and increases the blood volume.

Biochem/physiol Actions

Fludrocortisone acetate is a synthetic corticosteroid with more mineralocorticoid than glucocorticoid activity.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G G Krishna et al.
Kidney international, 43(5), 1097-1103 (1993-05-01)
Potassium depletion induced by dietary potassium restriction causes sodium retention while potassium supplementation augments urinary sodium excretion. The role of external potassium balance in modulating mineralocorticoid-induced sodium retention in humans is unknown. Accordingly, eight healthy subjects were studied at the
Autonomic Neurology (2014)
Stéphane Gibaud et al.
International journal of pharmaceutics, 269(2), 491-508 (2004-01-07)
Substitutive hormonal therapies have to be administered for long periods. Thus, the development of sustained-release forms, as microparticle suspensions, is interesting in order to improve patient compliance by reducing dosing frequencies and side effects. The aim of this work was
Ichiro Nakagawa et al.
Cerebrovascular diseases (Basel, Switzerland), 35(2), 131-137 (2013-02-15)
Hyponatremia is a common complication occurring in one third of patients after subarachnoid hemorrhage (SAH). One mechanism that likely mediates the development of hyponatremia in SAH is cerebral salt wasting syndrome (CSWS), which induces natriuresis and reduces total blood volume
R Taniguchi et al.
The Tohoku journal of experimental medicine, 186(3), 215-223 (1999-05-29)
It is known that some patients with primary aldosteronism show postoperative hyperkalemia, which is due to inability of the adrenal gland to secrete sufficient amounts of aldosterone. However, hyperkalemia is generally neither severe nor prolonged, in which replacement therapy with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service