Skip to Content
Merck
All Photos(1)

Key Documents

B7777

Sigma-Aldrich

Budesonide

≥99%

Synonym(s):

16,17-Butylidenebis(oxy)-11,21-dihydroxypregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H34O6
CAS Number:
Molecular Weight:
430.53
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99%

form

powder

originator

AstraZeneca

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@H]5OC(CCC)O[C@@]45C(=O)CO

InChI

1S/C25H34O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,16-18,20-22,26,28H,4-7,11-13H2,1-3H3/t16-,17-,18-,20+,21?,22+,23-,24-,25+/m0/s1

InChI key

VOVIALXJUBGFJZ-KWVAZRHASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Budesonide is a second generation glucocorticoid with low systemic absorption. It is used as an anti-inflammatory agent in the treatment of asthma, rhinitis, and inflammatory bowel disease. It inhibits the expression of chemokine mRNA and production of eotaxin and RANTES protein in primary human bronchial epithelial cells. Budesonide is currently in clinical trials for the prevention of lung cancer. It shows inhibitory effects on benzo[a]pyrene-induced carcinogenesis of the lung in mice.
Second generation glucocorticoid with low systemic absorption; anti-inflammatory agent; inhibits the expression of chemokine mRNA and production of eotaxin and RANTES protein.

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Repr. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Adrenal gland

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Bergeron et al.
American journal of respiratory and critical care medicine, 191(11), 1242-1249 (2015-04-04)
Systemic steroids are the standard treatment for bronchiolitis obliterans syndrome (BOS) after allogeneic hematopoietic stem cell transplantation (HSCT) despite their poor efficacy and disabling side effects. To evaluate the effectiveness and tolerance of budesonide/formoterol as an alternative treatment for BOS
Sissie Wong et al.
Respiratory research, 10, 72-72 (2009-08-05)
There is currently a vast amount of evidence in the literature suggesting that matrix metalloproteinases (MMPs) and tissue inhibitors of metalloproteinases (TIMPs) are involved in the pathogenesis of inflammatory airways diseases, such as asthma and COPD. Despite this, the majority
James Fingleton et al.
The Journal of allergy and clinical immunology, 136(3), 601-609 (2015-03-10)
Asthma and chronic obstructive pulmonary disease (COPD) are heterogeneous disorders encompassing different phenotypes of airflow obstruction, which might differ in their response to treatment. The aim of this study was to determine distinct phenotypes comprising the syndromes of asthma and
Koremu K Meja et al.
American journal of respiratory cell and molecular biology, 39(3), 312-323 (2008-04-19)
Oxidative stress as a result of cigarette smoking is an important etiologic factor in the pathogenesis of chronic obstructive pulmonary disease (COPD), a chronic steroid-insensitive inflammatory disease of the airways. Histone deacetylase-2 (HDAC2), a critical component of the corticosteroid anti-inflammatory
Marek Woynarowski et al.
The Journal of pediatrics, 163(5), 1347-1353 (2013-07-03)
To compare the effect of budesonide vs prednisone therapy both in combination with azathioprine in pediatric patients with autoimmune hepatitis (AIH). Forty-six patients with AIH (11 males and 35 females) aged 9-17 years were enrolled in a 6-month, prospective, double-blind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service