Skip to Content
Merck
All Photos(1)

Key Documents

F9428

Sigma-Aldrich

Fluticasone propionate

≥98% (HPLC), powder

Synonym(s):

(6α,11β,16α,17α)-6,9-Difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)androsta-1,4-diene-17-carbothioic acid S-(fluoromethyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H31F3O5S
CAS Number:
Molecular Weight:
500.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: ≥10 mg/mL

originator

GlaxoSmithKline

SMILES string

CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SCF

InChI

1S/C25H31F3O5S/c1-5-20(31)33-25(21(32)34-12-26)13(2)8-15-16-10-18(27)17-9-14(29)6-7-22(17,3)24(16,28)19(30)11-23(15,25)4/h6-7,9,13,15-16,18-19,30H,5,8,10-12H2,1-4H3/t13-,15+,16+,18+,19+,22+,23+,24+,25+/m1/s1

InChI key

WMWTYOKRWGGJOA-CENSZEJFSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Fluticasone blocks mast cells, eosinophils, lymphocytes, neutrophils and macrophages.

Biochem/physiol Actions

Fluticasone propionate is a second generation glucocorticoid. Used as an anti-inflammatory agent for asthma. Shown to enhance eosinophil apoptosis in a concentration-dependent manner via the glucocorticoid receptor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Endocrine system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Small Animal Clinical Pharmacology (2008)
Kate McKeage
Drugs, 73(2), 195-206 (2013-02-12)
The corticosteroid fluticasone propionate (fluticasone) and the long-acting β₂-adrenoceptor agonist formoterol fumarate (formoterol) have been combined in a single, pressurized, metered-dose, aerosol inhaler for the maintenance treatment of patients aged ≥12 years with persistent asthma. This article reviews the clinical
Melanie Manetsch et al.
PloS one, 8(3), e59635-e59635 (2013-03-28)
Mitogen-activated protein kinase phosphatase 1 (MKP-1) represses MAPK-driven signalling and plays an important anti-inflammatory role in asthma and airway remodelling. Although MKP-1 is corticosteroid-responsive and increased by cAMP-mediated signalling, the upregulation of this critical anti-inflammatory protein by long-acting β2-agonists and
David Price et al.
The Journal of allergy and clinical immunology, 131(3), 763-771 (2013-02-06)
Many asthmatic patients are unable to quit cigarettes; therefore information is needed on treatment options for smokers. This study evaluates 10 mg/d montelukast and 250 μg of fluticasone propionate twice daily, each compared with placebo, in patients with self-reported active smoking
Fouad J Moawad et al.
The American journal of gastroenterology, 108(3), 366-372 (2013-02-13)
Patients with clinical symptoms of esophageal dysfunction and dense eosinophilic infiltration of the esophageal mucosa are suspected to have eosinophilic esophagitis (EoE). Topical steroids are often used as first-line therapy for EoE, although some patients respond clinically to proton pump

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service