Skip to Content
Merck

Skip To

F3290

FLAG® Peptide

≥85% (HPLC), lyophilized powder

Synonym(s):

Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys, ddddk peptide, dykddddk peptide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C41H60N10O20
Molecular Weight:
1012.97
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

FLAG® Peptide, lyophilized powder

assay

≥85% (HPLC)

Quality Level

form

lyophilized powder

mol wt

1012.97 Da

shipped in

wet ice

storage temp.

2-8°C

SMILES string

NCCCC[C@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](N)CC(O)=O)C(O)=O

InChI

1S/C41H60N10O20/c42-11-3-1-5-22(45-36(65)24(13-19-7-9-20(52)10-8-19)47-34(63)21(44)14-29(53)54)35(64)48-26(16-31(57)58)38(67)50-28(18-33(61)62)40(69)51-27(17-32(59)60)39(68)49-25(15-30(55)56)37(66)46-23(41(70)71)6-2-4-12-43/h7-10,21-28,52H,1-6,11-18,42-44H2,(H,45,65)(H,46,66)(H,47,63)(H,48,64)(H,49,68)(H,50,67)(H,51,69)(H,53,54)(H,55,56)(H,57,58)(H,59,60)(H,61,62)(H,70,71)/t21-,22-,23-,24-,25-,26-,27-,28-/m0/s1

InChI key

XZWYTXMRWQJBGX-VXBMVYAYSA-N

General description

The FLAG peptide is used for the competitive elution of amino-terminal, Met-amino-terminal or carboxy-terminal FLAG fusion proteins from the Anti-FLAG M1 or Anti-FLAG M2 antibody (in solution or bound to an agarose gel).

Application

Gently elutes FLAG fusion proteins from ANTI-FLAG® M1 and M2 affinity resins. Usual working concentration is 100 μg/ml. Will not elute 3X FLAG fusion proteins. For this application, use 3X FLAG Peptide, F4799.

Learn more product details in our FLAG® application portal.

Preparation Note

For stock solution, dissolve in TBS (10mM Tris HCL, 150 mM NaCl, pH7.4) to a final concentration of 5 mg/mL.

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F4799SAE0194I2149
assay

≥85% (HPLC)

assay

≥90% (HPLC/MS)

assay

≥95% (HPLC)

assay

≥97% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

powder

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

shipped in

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

mol wt

1012.97 Da

mol wt

-

mol wt

-

mol wt

1102.15 Da


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Omar H Vandal et al.
Nature medicine, 14(8), 849-854 (2008-07-22)
Acidification of the phagosome is considered to be a major mechanism used by macrophages against bacteria, including Mycobacterium tuberculosis (Mtb). Mtb blocks phagosome acidification, but interferon-gamma (IFN-gamma) restores acidification and confers antimycobacterial activity. Nonetheless, it remains unclear whether acid kills
Barbara G Mellone et al.
PLoS genetics, 7(5), e1002068-e1002068 (2011-05-19)
Semi-conservative segregation of nucleosomes to sister chromatids during DNA replication creates gaps that must be filled by new nucleosome assembly. We analyzed the cell-cycle timing of centromeric chromatin assembly in Drosophila, which contains the H3 variant CID (CENP-A in humans)
Jingxiang Huang et al.
Cancer research, 69(15), 6107-6114 (2009-07-16)
Mutations in the TSC1 and TSC2 tumor suppressor genes give rise to the neoplastic disorders tuberous sclerosis complex (TSC) and lymphangioleiomyomatosis. Their gene products form a complex that is a critical negative regulator of mammalian target of rapamycin (mTOR) complex



Global Trade Item Number

SKUGTIN
F3290-25MG04061835573004
F3290-4MG04061833613689

Questions

Reviews

No rating value

Active Filters