ER0100
Endoplasmic Reticulum Isolation Kit
isolation of intact ER from mammalian soft tissues and cultured cells
Synonym(s):
ER isolation kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
The endoplasmic reticulum (ER) is a network of interconnected tubules, vesicles, and sacs. It plays a role in specialized cellular functions including protein synthesis, sequestering of calcium, production of steroids, storage and production of glycogen, and insertion of membrane proteins.
Application
The Endoplasmic Reticulum Isolation Kit contains all the reagents required for ER preparations with various degrees of purity: crude ER (microsomes), Ca2+ precipitated rough ER (RER) enriched microsomes, and density gradient purified rough ER (RER) and smooth ER (SER). The ER isolation process can be monitored by measuring the activity of NADPH cytochrome c reductase using the Cytochrome c Reductase Assay Kit, (Cat. No. CY0100). The enzyme is an ER membrane protein that is commonly used as an ER marker. The isolated ER can be used for studying the cytochrome P450 system and xenobiotic metabolism, for studying lipid metabolism, and for the recovery of ER membrane and lumenal proteins. The kit was tested with rat liver, kidney, and brain; mouse liver; rabbit liver, kidney, spleen, heart, and brain; and with Jurkat and HeLa cell lines.
The Endoplasmic Reticulum Isolation Kit is used for the efficient enrichment of functional endoplasmic reticulum (ER) from mammalian tissue or cell culture. The isolated ER are useful for protein synthesis, degradation and metabolic pathway analysis. Metabolic abnormalities of ER include carnitine acetyltransferase deficiency in children, cystic fibrosis and many more ER protein retention disorders. Suitable for up to 50 g of tissue or ~20 mL of packed tissue.
Features and Benefits
- Specially formulated extraction reagents for research scale applications - save time and minimize waste
- Includes a calcium chloride solution - for quick and simple precipitation of rough ER without need for ultracentrifugation
- Produces functional, intact organelles - resulting ER are suitable for functional studies, lipid metabolism, and protein profiling
- Compatible with products for structure confirmation - easily confirm intactness with companion kit test for cytochrome C reductase (CY0100)
- Can be used to isolate ER from soft animal tissues and cultured cells
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cancer discovery, 9(3), 396-415 (2018-12-20)
Resistance to BRAF and MEK inhibitors (BRAFi + MEKi) in BRAF-mutant tumors occurs through heterogeneous mechanisms, including ERK reactivation and autophagy. Little is known about the mechanisms by which ERK reactivation or autophagy is induced by BRAFi + MEKi. Here
Nature communications, 10(1), 3391-3391 (2019-07-31)
Non-alcoholic fatty liver disease (NAFLD) is the hepatic manifestation of the metabolic syndrome that elevates the risk of hepatocellular carcinoma (HCC). Although alteration of lipid metabolism has been increasingly recognized as a hallmark of cancer cells, the deregulated metabolic modulation
Nature communications, 7, 10880-10880 (2016-03-17)
Membrane transporters are key determinants of therapeutic outcomes. They regulate systemic and cellular drug levels influencing efficacy as well as toxicities. Here we report a unique phosphorylation-dependent interaction between drug transporters and tyrosine kinase inhibitors (TKIs), which has uncovered widespread
Nature, 551(7679), 198-203 (2017-11-10)
The rate of behavioural decline in the ageing population is remarkably variable among individuals. Despite the considerable interest in studying natural variation in ageing rate to identify factors that control healthy ageing, no such factor has yet been found. Here
The Journal of biological chemistry, 284(3), 1410-1418 (2008-11-20)
Glycoprotein Ib alpha (GpIbalpha), a trans-membrane glycoprotein, is expressed on the surface of megakaryocytes and platelets, where, in association with glycoprotein Ib beta, glycoprotein V, and glycoprotein IX, it normally forms the von Willebrand factor receptor (vWFR). A fully functional
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service