Skip to Content
Merck
All Photos(1)

Key Documents

CPISO

Sigma-Aldrich

Chloroplast Isolation Kit

isolation of intact chloroplasts from leaves

Synonym(s):

Chloroplast Purification Kit, Isolation Kit for Chloroplast, Kit for Chloroplast Isolation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

Quality Level

technique(s)

centrifugation: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

The Chloroplast Isolation Kit provides an efficient procedure for isolating intact chloroplasts from plant leaves. The procedure involves mechanical cell wall and cell membrane breakage, removal of cell debris and leaf tissue by filtration, collection of total cell chloroplast fraction by centrifugation, and separation of intact chloroplasts from broken chloroplasts using a Percoll® layer or gradient.

Application

Chloroplast Isolation Kit may be used in chloroplast organelle research to isolate intact chloroplasts from plant leaves for structural and functional studies.

Features and Benefits

  • Specifically formulated buffer - Saves time using easy to follow simple and proven protocols
  • Isolates intact chloroplasts - Useful for studies of such processes as carbon assimilation, electron flow and phosphorylation, metabolic transport or protein targeting
  • Total organelle protein complement - Useful for functional studies, metabolic transport, proteomic profiling, and genomic applications
  • Convenient format - Saves time and reduces waste

Suitability

The Chloroplast Isolation Kit has been tested for use with spinach, pea, lettuce, cabbage, mangold, and tobacco.

Kit Components Also Available Separately

Product No.
Description
SDS

  • Chloroplast Isolation Buffer 5x (CIB) 500 mL

  • A8022Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96% 3 gSDS

  • Chloroplast Isolation Buffer 5x (CIB) 4 ea

  • P4937Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture 100 mLSDS

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoli Luo et al.
PloS one, 8(1), e54002-e54002 (2013-01-22)
In plants, CuZn superoxide dismutase (CuZnSOD, EC l.15.1.1), ascorbate peroxidase (APX, EC 1.11.1.11), and catalase (CAT, EC l.11.1.6) are important scavengers of reactive oxygen species (ROS) to protect the cell from damage. In the present study, we isolated three homologous
Esau Megias et al.
The protein journal, 37(3), 290-299 (2018-05-29)
Tymovirus is a genus of plant pathogenic viruses that infects several dicotyledonous plants worldwide, causing serious diseases in economically important crops. The known cytopathic effect on the host cell organelles involves chloroplast membrane deformation and the induction of vesicles in
Yanzhong Luo et al.
Frontiers in plant science, 11, 613-613 (2020-06-13)
The transportation of proteins encoded by nuclear genes from plant cytosol to chloroplast is essential for chloroplast functions. Proteins that have a chloroplast transit peptide (cTP) are imported into chloroplasts via translocases on the outer and inner chloroplast envelope. How
Martha Sainz et al.
Physiologia plantarum, 140(1), 46-56 (2010-05-22)
Drought and heat stress have been studied extensively in plants, but most reports involve analysis of response to only one of these stresses. Studies in which both stresses were studied in combination have less commonly been reported. We report the
The Complete Chloroplast Genome of 17 Individuals of Pest Species <I>Jacobaea vulgaris</I>: SNPs, Microsatellites and Barcoding Markers for Population and Phylogenetic Studies.
Doorduin, L., et al.
DNA Res., 1-13 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service