It is possible that the nuclear and cytoplasmic extracts can be used for a Western blot. The kit is designed to maintain the functional activity of proteins for assays such as EMSAs (with non-denaturing PAGE) where the protein structure should remain relatively intact. For Western blots, proteins are linearized using denaturing PAGE gels, typically with the presence of SDS detergent or other denaturants. To proceed, it is suggested to use RIPA buffer as the extraction buffer/dilution buffer for each fraction separately after separating the cytoplasmic fraction from the nuclear fraction.
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Level
usage
1 kit sufficient for 10 extractions (1 ml packed cell volume), 1 kit sufficient for 100 extractions (100 μl packed cell volume)
technique(s)
protein extraction: suitable, western blot: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Application
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) protein extraction: suitable, western blot: suitable | technique(s) - | technique(s) ChIP: suitable | technique(s) - |
| usage kit sufficient for 10 extractions (1 ml packed cell volume), kit sufficient for 100 extractions (100 μl packed cell volume) | usage sufficient for 25 nuclei preparations (~1-10×107 cells/preparation) | usage kit sufficient for 30 extractions (20 g of fresh or frozen leaves) | usage - |
| Quality Level 300 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| shipped in dry ice | shipped in wet ice | shipped in wet ice | shipped in - |
| storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C |
Still not finding the right product?
Explore all of our products under NuCLEAR™ Extraction Kit
Kit Components Also Available Separately
signalword
Danger
Storage Class
8A - Combustible corrosive hazardous materials
flash_point_f
188.6 °F - closed cup
flash_point_c
87 °C - closed cup
wgk
WGK 3
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| NXTRACT-1KT | 04061838256133 |
-
Is it possible to use the cytoplasmic and nuclear fractions for Western Blotting?
1 answer-
Helpful?
-
-
Can Product NXTRACT, CelLytic™ NuCLEAR™ Extraction Kit, be used to make nuclear protein extracts for EMSAs/DNA binding studies?
1 answer-
Several references describe the use of the N-XTRACT kit in sample preparation for EMSA work.1. Xie, J., et al., β2-Microglobulin as a negative regulator of the immune system: high concentrations of the protein inhibit in vitro generation of functional dendritic cells. Blood, 101(10), 4005-4012 (2003).2. Marambaud, P., et al., A CBP Binding Transcriptional Repressor Produced by the PS1/ε-Cleavage of N-Cadherin Is Inhibitedby PS1 FAD Mutations. Cell, 114, 635-645 (2003).3. Chen, G., et al., Phosphorylated FADD induces NF-κB, perturbs cell cycle, and is associated with poor outcome in lung adenocarcinomas. Proc. Nat. Acad. Sci. USA, 102(35), 12507-12512 (2005).
Helpful?
-
-
What are the options for lysis of my cells to make extracts when using Product NXTRACT, CelLytic™ NuCLEAR™ Extraction Kit?
1 answer-
The use of detergent will lead to maximum protein yield. However we find that the hypotonic lysis is extremely effective and in addition for very fragile cells we offer an isotonic lysis option.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
When using Product NXTRACT, CelLytic™ NuCLEAR™ Extraction Kit, what can I use for the mechanical lysis of my cells?
1 answer-
We recommend the use of a glass tissue homogenizer, with a type B pestle, or alternatively a syringe with a narrow-gauge (No. 27) hypodermic needle may be used.
Helpful?
-
-
How "pure" is the nuclear extraction using Product NXTRACT, CelLytic™ NuCLEAR™ Extraction Kit?
1 answer-
It is important to keep in mind that this kit will provide a crude nuclear fraction. Depending on the care taken at the step where the nuclei are pelleted and the supernatant containing the cytoplasmic fraction is removed, we have found that the cytoplasmic contamination of the nuclear pellet may be a few percent.
Helpful?
-
-
Is Product NXTRACT, CelLytic™ NuCLEAR™ Extraction Kit, suitable for use on tissues?
1 answer-
We have used the kit with several tissues: Mouse lung and brain, Rat liver and Rabbit muscle. For these tissues we usually used the kit with 100 mg tissue. After extraction with 140 μl Extraction buffer, we obtained an average yield of protein concentration as follows: 3.2 mg/ml for brain, 4.1 mg/ml for lung, 3.3 mg/ml for muscle and 10 mg/ml for liver.
Helpful?
-





