EP0030113527
Eppendorf® Deepwell 96 Well PCR Plate with SafeCode System (data matrix code, barcode and plain text labels)
2000 µL per well, clear white wells, PCR clean
All Photos(1)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
clear white wells
polypropylene
polypropylene
size:
96 well
sterility:
non-sterile
feature:
PCR clean
lid: no
lid: no
Recommended Products
material
clear white wells
polypropylene
sterility
non-sterile
feature
PCR clean
lid: no
packaging
pkg of 20 plates (5 bags × 4 plates)
manufacturer/tradename
Eppendorf® 0030113527
size
96 well
well size
2000 μL , per well
suitability
suitable for PCR
General description
Deepwell Plate 96/2000 µL, wells clear, PCR clean, white, with 2D SafeCode (DataMatrix/ECC200) and human readable code, 20 plates (5 bags x 4 plates)
Features and Benefits
- 3-level coding for safe sample and vessel identification
- Codes on three sides of plates for flexible reading
- ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
- Additional, ID-specific information on plates available via Eppendorf DataPort for documentation purposes
- Direct import into eLabInventory and eLabJournal software for convenient handling
- Import to other database systems possible
- Manufactured from high-quality polypropylene for highest sample purity
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)