Key Documents
EP0030113543
Eppendorf® Deepwell 96 Well PCR Plate with SafeCode System (data matrix code, barcode and plain text labels)
500 µL per well, clear white wells, PCR clean
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material
clear white wells
polypropylene
sterility
non-sterile
feature
PCR clean
lid: no
packaging
pkg of 40 plates (5 bags × 8 plates)
manufacturer/tradename
Eppendorf® 0030113543
well size
500 μL , per well
suitability
suitable for PCR
General description
Deepwell Plate 96/500 µL, wells clear, PCR clean, white, with 2D SafeCode (DataMatrix/ECC200) and human readable code, 40 plates (5 bags x 8 plates)
Features and Benefits
  • 3-level coding for safe sample and vessel identification
  • Codes on three sides of plates for flexible reading
  • ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Additional, ID-specific information on plates available via Eppendorf DataPort for documentation purposes
  • Direct import into eLabInventory and eLabJournal software for convenient handling
  • Import to other database systems possible
  • Manufactured from high-quality polypropylene for highest sample purity
Legal Information
Eppendorf is a registered trademark of Eppendorf AG