Skip to Content
Merck
All Photos(3)

Key Documents

E2761

Sigma-Aldrich

ExtrAvidin®−FITC

buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

FITC conjugate

form

buffered aqueous solution

extent of labeling

3-6 mol fluorochrome per mol protein

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 20 μg/mL using human tissues
indirect immunofluorescence: 1:200

shipped in

dry ice

storage temp.

−20°C

General description

Avidin is a tetrameric or dimeric biotin-binding protein produced in the oviducts of birds, reptiles and amphibians and deposited in the whites of their eggs. It consists of four high affinity binding sites for biotin. ExtrAvidin is prepared from egg white avidin. It is a modified form of affinity purified avidin and combines high specific activity of avidin with the low background staining of streptavidin. It is a biotin binding protein produced by the bacteria Streptomyces avidinii. ExtrAvidin has been conjugated to fluorescein isothiocyanate (FITC) and is used for various techniques.

Application

ExtrAvidin®-FITC is suitable for the following applications:
  • immunohistochemistry (formalin-fixed, paraffin-embedded sections) at a concentration of 20μg/mL using human tissues.
  • Immunofluorescence
  • competitive binding assay

Physical form

Solution in 0.01 M phosphate buffered saline, containing 10% (v/v) 0.5 M carbonate buffer, pH 9.5, and 15 mM sodium azide

Preparation Note

Affinity purified protein

Legal Information

ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therapeutic monoclonal antibodies for Ebola virus infection derived from vaccinated humans
Rijal P, et al.
Cell Reports, 27, 172-186 (2019)
Xueying Wang et al.
eLife, 3, e03427-e03427 (2014-07-17)
To understand the neural origins of rhythmic behavior one must characterize the central pattern generator circuit and quantify the population size needed to sustain functionality. Breathing-related interneurons of the brainstem pre-Bötzinger complex (preBötC) that putatively comprise the core respiratory rhythm
D R Hampson et al.
The Journal of biological chemistry, 274(47), 33488-33495 (1999-11-24)
Metabotropic glutamate receptors (mGluRs) are G-protein-coupled glutamate receptors that subserve a number of diverse functions in the central nervous system. The large extracellular amino-terminal domains (ATDs) of mGluRs are homologous to the periplasmic binding proteins in bacteria. In this study
J L Tremoleda et al.
European journal of nuclear medicine and molecular imaging, 43(9), 1710-1722 (2016-05-08)
Traumatic spinal cord injury (SCI) is a devastating condition which affects millions of people worldwide causing major disability and substantial socioeconomic burden. There are currently no effective treatments. Modulating the neuroinflammatory (NI) response after SCI has evolved as a major
R S Schwartz et al.
The Biochemical journal, 327 ( Pt 2), 609-616 (1997-11-14)
We report the cloning and sequencing from human reticulocytes of cDNA coding for the Cl- channel-associated protein, pICln. Human reticulocyte pICln (HRpICln) cDNA encodes a protein (predicted molecular mass 26293Da) identical with human non-pigmented ciliary epithelial cell pICln. By using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service