Skip to Content
Merck
All Photos(1)

Documents

385911

Sigma-Aldrich

Hyaluronic Acid Binding Protein, Bovine Nasal Cartilage, Biotinylated

Synonym(s):

Binding Protein for Hyaluronic Acid, Biotinylated Hyaluronic Acid Protein, Bovine Nasal Cartilage Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.21

biological source

bovine (nasal cartilage)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

General description

Affinity purified. Binds specifically and strongly to hyaluronic acid (≥ 2000 M.W.). Composed of two binding polypeptides, which are derived from N-terminal regions of the hyaluronic acid binding proteoglycan and linkage proteins. Useful for ELISA and histochemistry.
Affinity-purified. Binds specifically and strongly to hyaluronic acid (≥2000 M.W.). Composed of two binding polypeptides, which are derived from N-terminal regions of hyaluronic acid binding proteoglycan and linkage proteins. Useful for ELISA and histochemistry.
M.W. 70,000-80,000 and 40,000-45,000.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.1 M sodium bicarbonate solution, pH 8.3

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Reconstitute with 100 µl distilled H₂O.

Other Notes

Tammi, R., et al. 1998. J. Biol. Chem. 273, 28878.
Kohda, D., et al. 1996. Cell 86, 767.
Heinegard, D., and Hascall, V.C. 1974. J. Biol. Chem. 249, 4250.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Appeltant et al.
Reproduction & fertility, 2(4), L10-L12 (2022-02-05)
To visualise tissues to determine the presence of disease or simply to understand anatomy, it is important to preserve fresh tissue. Fixatives are chemical solutions that preserve tissues to enable microscopic evaluation. However, some fixatives introduce artefact such as shrinkage
Marie C Hasselluhn et al.
Cells, 9(1) (2019-12-28)
The KrasG12D/+;LSL-Trp53R172H/+;Pdx-1-Cre (KPC) mouse model is frequently employed for preclinical therapeutic testing, in particular in regard to antistromal therapies. Here, we investigate the prognostic implications of histopathological features that may guide preclinical trial design. Pancreatic tumor tissue from n =
James E Parkinson et al.
Immunology and cell biology, 99(6), 640-655 (2021-02-16)
Allergic airway inflammation is heterogeneous with variability in immune phenotypes observed across asthmatic patients. Inflammation has been thought to directly contribute to airway remodeling in asthma, but clinical data suggest that neutralizing type 2 cytokines does not necessarily alter disease
Lisa DiNatale et al.
Frontiers in medicine, 8, 724344-724344 (2021-10-05)
Topical antiaging products are often a first-line intervention to counter visible signs of facial photoaging, aiming for sustained cosmetic improvement. However, prolonged application of a single active topical compound was observed clinically to lead to a plateau effect in improving
Kazadi Nadine Mutoji et al.
BMC immunology, 22(1), 52-52 (2021-08-06)
Current research suggests that the glial scar surrounding penetrating brain injuries is instrumental in preserving the surrounding uninjured tissue by limiting the inflammatory response to the injury site. We recently showed that tumor necrosis factor (TNF)-stimulated gene-6 (TSG-6), a well-established

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service