Skip to Content
Merck
All Photos(3)

Key Documents

E2636

Sigma-Aldrich

ExtrAvidin® –Alkaline Phosphatase

buffered aqueous solution

Synonym(s):

Alkaline Phosphatase Reagent, ExtrAvidin Alkaline Phosphatase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

alkaline phosphatase conjugate

form

buffered aqueous solution

technique(s)

dot blot: 1:300,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100 using human tissues sections
indirect ELISA: 1:60,000

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Avidin is a tetrameric or dimeric biotin-binding protein produced in the oviducts of birds, reptiles and amphibians and deposited in the whites of their eggs. It consists of four high affinity binding sites for biotin. ExtrAvidin® is prepared from egg white avidin. It is a modified form of affinity purified avidin with high specific activity of avidin and low background staining of streptavidin. It is a biotin binding protein produced by the bacteria Streptomyces avidinii. ExtrAvidin has been conjugated to Alkaline Phosphatase and is used for various techniques.

Application

ExtrAvidin® -Alkaline Phosphatase has been used in:
  • Dot blot
  • Immunohistochemistry (formalin-fixed, paraffin-embedded sections)
  • Indirect ELISA
  • Enzyme linked immunosorbent assay (ELISA)
  • Enzyme-linked immunosorbent spot (ELISPOT) analysis
  • Competitive ELISA

Biochem/physiol Actions

Alkaline Phosphatase catalyzes the hydrolysis of esters to yield phosphoric acid. It is also facilitate the transport of substances across membranes.

Physical form

Solution in 0.05 M Tris-HCl buffer, pH 8.0, containing 1 mM MgCl2, 1% bovine serum albumin and 15 mM sodium azide

Preparation Note

Affinity purified protein

Legal Information

ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W E Sweeney et al.
Kidney international, 56(2), 406-413 (1999-08-05)
Recessively transmitted polycystic kidney disease (PKD) in many murine models is characterized by the initial formation of proximal tubular cysts (stage 1), followed by growth and enlargement of renal collecting tubule (CT) cysts (stage 2). Previous studies have reported that
Joanne L Maki et al.
Clinical and diagnostic laboratory immunology, 10(5), 876-881 (2003-09-11)
Fish acquire protective immunity against the ciliated protozoan parasite Ichthyophthirius multifiliis following sublethal infection or inoculation with I. multifiliis immobilization antigens (i-antigens). In both cases, parasite-immobilizing antibodies have been identified in sera and mucosal secretions. To investigate the kinetics of
C E Rutten et al.
Leukemia, 22(7), 1387-1394 (2008-04-18)
Mismatching for human leukocyte antigen (HLA)-DPB1 in unrelated donor hematopoietic stem cell transplantation (URD-SCT) has been associated with a decreased risk of disease relapse, indicating that HLA-DP may represent a target for graft-versus-leukemia (GVL) reactivity in HLA class II-expressing hematological
Alkaline Phosphatase (2103)
Jenny Meegan et al.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 22(6), 856-862 (2010-11-23)
A competitive enzyme-linked immunosorbent assay (cELISA) was developed by using a whole-cell antigen from a marine Brucella sp. isolated from a harbor seal (Phoca vitulina). The assay was designed to screen sera from multiple marine mammal species for the presence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service