Skip to Content
Merck
All Photos(3)

Key Documents

E0258

Sigma-Aldrich

Elastase from porcine pancreas

Type IV, Protein 50-90 %, lyophilized powder, ≥4.0 units/mg protein (biuret)

Synonym(s):

Elastase from hog pancreas, Pancreatopeptidase E

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Porcine pancreas

type

Type IV

form

lyophilized powder

specific activity

≥4.0 units/mg protein (biuret)

composition

Protein, 50-90%

foreign activity

trypsin ≤50 BAEE units/mg protein

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Elastase is a single polypeptide chain of 240 amino acid residues and contains four disulfide bridges. The molecular mass is approximately 25.9 kDa. The enzyme is synthesized as an inactive zymogen, proelastase, which is converted to the active form by limited proteolysis at the N-terminal by trypsin.

Application

Elastase from Sigma has been used to examine the extent of proteolytic degradation of BSA that is treated with hydroxyl radical. It has also been used to purify elastase-inhibitory lipid derivative from a cyanobacterium, Microcystis Ku2.
Elastase from porcine pancreas has been used in a study to assess the molecular bases for human leucocyte elastase inhibition. Elastase from porcine pancreas has also been used in a study to investigate the molecular cloning and expression of serum calcium-decreasing factor (caldecrin).
Elastase from porcine pancreas has been used:
  • to treat vero cells to study its effects on syncytium formation
  • as a positive control in protease assays
  • as a component in RPMI 1640 to isolate human aortic smooth muscle cells (HASMCs) from the aortic tissue

Biochem/physiol Actions

Elastase hydrolyses elastin, the specific protein of elastic fibers, and digests hemoglobin, casein and fibrin.
Elastase is a serine protease with broad specificity as it cleaves protein at the carboxyl side of small hydrophobic amino acids such as Ile, Gly, Ala, Ser, Val and Leu. The enzyme also hydrolyzes amides and esters such as N-Benzoyl-L-alanine methyl ester. The pH optimum is found to be 8.0-8.5. It does not require any activator, but it is inhibited by diisopropyl fluorophosphate, phenylmethanesulfonyl fluoride, α2-macroglobulin, α1-antitrypsin, sulfonyl fluorides and p-dinitrophenyl diethylphosphate and high salt concentrations. It is extensively used in tissue and cell dissociation procedures. Elastase is effective in the isolation of Type II lung cells. Elastase hydrolyses elastin, the specific protein of elastic fibers, and digests hemoglobin, casein and fibrin.
Elastase is a serine protease with broad specificity as it cleaves protein at the carboxyl side of small hydrophobic amino acids such as Ile, Gly, Ala, Ser, Val, and Leu. The enzyme also hydrolyzes amides and esters such as N-Benzoyl-L-alanine methyl ester. The pH optimum is found to be 8.0-8.5. It does not require any activator, but it is inhibited by diisopropyl fluorophosphate, phenylmethanesulfonyl fluoride, α2-macroglobulin, α1-antitrypsin, sulfonyl fluorides and p-dinitrophenyl diethylphosphate and high salt concentrations. It is extensively used in tissue and cell dissociation procedures. Elastase is effective in the isolation of Type II lung cells.

Packaging

Package size based on protein content

Unit Definition

One unit will hydrolyze 1.0 μmole of N-succinyl-L-Ala-Ala-Ala-p-nitroanilide per min, pH 8.0 at 25 °C.

Physical form

Contains sodium carbonate.

Preparation Note

A further purification of Type III, E 0127, by affinity chromatography to reduce trypsin activity

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suvendra Nath Bagchi et al.
Indian journal of microbiology, 51(4), 496-500 (2012-10-02)
A unicyanobacterial isolate of cyanobacterium, identified as Microcystis Ku2, produced a mammalian elastase-inhibitory lipid derivative. Protease inhibitors in cyanobacteria are unequivocally peptides. Since this metabolite appeared in lipid phase, we worked on a hypothesis that whether metabolite other than peptides
Digestive enzymes and stylet morphology of Deraeocoris nebulosus (Hemiptera: Miridae), a predacious plant bug
Boyd DW, et al.
Annals of the Entomological Society of America, 395-401 (2002)
Regulation of human aortic endothelial cell-derived mesenchymal growth factors by allogeneic lymphocytes in vitro. A potential mechanism for cardiac allograft vasculopathy.
Wagner CR, et al.
The Journal of Clinical Investigation, 1269-1277 (1993)
N Rugani et al.
Biochimica et biophysica acta, 1247(2), 185-194 (1995-03-15)
Porcine colipase, the protein cofactor of pancreatic lipase, was isolated from pancreas freshly collected on animals and from a side fraction from the production of insulin (Novo Nordisk A/S). Samples of purified colipase were analyzed for homogeneity by polyacrylamide gel
Cloning and overexpression of extracellular elastase from Pseudomonas aeruginosa
Raftari M, et al.
European journal of inflammation, 11(1), 55-60 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service