Skip to Content
Merck
All Photos(8)

Key Documents

T3824

Sigma-Aldrich

Tyrosinase from mushroom

lyophilized powder, ≥1000 unit/mg solid

Synonym(s):

Catechol Oxidase, Monophenol Monooxygenase, Monophenol, dihydroxyphenylalanine:oxygen oxidoreductase, Polyphenol Oxidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

fungus (mushroom)

form

lyophilized powder

specific activity

≥1000 unit/mg solid

mol wt

119.5 kDa by electrophoresis
128 kDa by sedimentation velocity diffusion
133 kDa by LS

application(s)

diagnostic assay manufacturing

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Isoelectric point (pI): 4.7-5
pH optimum is 6-7

Molecular weight: 128 kDa by sedimentation velocity diffusion; 133 kDa by light-scattering measurements, and 119.5 kDa by electrophoresis.

Tyrosinase is a copper-containing oxidase, which has activity for both catechols and cresol. It is responsible for browning reactions. The enzyme is reported to have two binding sites for aromatic substrates and a different binding site for oxygen-copper. The copper is probably Cu(I), with inactivation involving oxidation to Cu(II) ion.
Tyrosinase is a copper-containing oxidase with activity for catechols and cresol. Tyrosinase participates in the conversion of tyrosine to dihydroxyphenylalanine (DOPA), and polymerization of DOPA leads to melanin formation. Tyrosinase has a role in browning reactions. The enzyme is said to have two binding sites for aromatic substrates and a distinct binding site for oxygen-copper.

Application

Tyrosinase activity has been assessed in a study that developed an alternative therapeutic agent for treating hyperpigmentation. Tyrosinase has also been used in a study to investigate the oculocutaneous albinism phenotype in the Pakistani population.

Biochem/physiol Actions

Converts tyrosine to L-DOPA or tyramine to dopamine

Physical properties

Isoelectric point (pI): 4.7-5
pH optimum is 6-7

Molecular weight: 128 kDa by sedimentation velocity diffusion; 133 kDa by light-scattering measurements, and 119.5 kDa by electrophoresis.

Unit Definition

One unit = ΔA280 of 0.001 per min at pH 6.5 at 25 °C in 3 mL reaction mix containing L-tyrosine.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyrosinase inhibitory effects and inhibition mechanisms of nobiletin and hesperidin from citrus peel crude extracts.
Zhang, et al.
Journal of Enzyme Inhibition and Medicinal Chemistry, 22, 83-90 (2019)
A T S Jorge et al.
International journal of cosmetic science, 34(5), 435-440 (2012-06-29)
Several treatments for skin whitening are available today, but few of them are completely adequate, especially owing to the carcinogenic potential attributed to classical drugs like hydroquinone, arbutin and kojic acid. To provide an alternative and safer technology for whitening
Shilpi Goenka et al.
Molecules (Basel, Switzerland), 25(16) (2020-08-13)
Previous studies have reported that estrogen hormone promotes melanogenesis while progesterone inhibits it. A selective estrogen receptor modulator (SERM), tamoxifen, has been shown to promote melanogenesis; however, to date, there have been no reports on the effects of a selective
Thomas J Jaworek et al.
Orphanet journal of rare diseases, 7, 44-44 (2012-06-28)
Oculocutaneous albinism (OCA) is caused by a group of genetically heterogeneous inherited defects that result in the loss of pigmentation in the eyes, skin and hair. Mutations in the TYR, OCA2, TYRP1 and SLC45A2 genes have been shown to cause
Rongchao Zhang et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 2582579-2582579 (2020-08-11)
Whitening cosmetics market has a bright future, and pure natural whitening products of traditional Chinese medicine have always been a research hotspot. In this research, the whitening active ingredient of Chinese medicine Trichosanthes pulp was isolated and purified for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service