D9692
Plant Protoplast Digest/Wash Solution
Rapid isolation of viable protoplasts from plant tissue
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105500
NACRES:
NA.56
Recommended Products
General description
The Plant Protoplast Digest/Wash Solution is formulated to facilitate the rapid isolation of viable protoplasts from plant tissue. Plant cells are surrounded by a rigid, semi-permeable cell wall composed primarily of three classes of polysaccharides: cellulose, hemicellulose, and pectin. The Plant Protoplast Digest/Wash Solution can be used for the digestion of the cell wall after the addition of enzymes that hydrolyze these polysaccharides, such as cellulase, pectinase, or pectolyase. The Plant Protoplast Digest/Wash Solution can be subsequently used to wash away any remaining hydrolytic enzymes after digestion is complete.
Plant protoplasts are typically used for any of a number of downstream applications. These applications include, but are not limited to, transient gene expression, viral transfection assays, somatic hybridization, electrophysiological studies, and morphological studies.
Plant protoplasts are typically used for any of a number of downstream applications. These applications include, but are not limited to, transient gene expression, viral transfection assays, somatic hybridization, electrophysiological studies, and morphological studies.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Akira Inoue et al.
Marine biotechnology (New York, N.Y.), 13(2), 256-263 (2010-04-16)
Functional recombinant abalone alginate lyase (rHdAly) and β-1,4-endoglucanase (rHdEG66) were expressed as secreted proteins with baculoviral expression systems. The specific activity of each recombinant enzyme, 2,490 and 18.2 U/mg for rHdAly and rHdEG66, respectively, was comparable to its native form
Shiu-Cheung Lung et al.
Plant cell reports, 30(4), 473-484 (2010-11-26)
Although transient gene expression using reporters such as green fluorescent protein is a versatile tool for examining gene functions and intracellular protein trafficking, the establishment of a highly efficient gene manipulation method remains a challenge in many plant species. A
Advanced patch-clamp techniques and single-channel analysis.
White, P. J. et al
The Journal of Experimental Biology, 50, 1037-1054 (1999)
The cell wall.
Carpita, N., and McCann, M. et al.
Biochemistry, 52-108 null
R Ma et al.
Plant cell reports, 22(5), 320-327 (2003-12-04)
A method for somatic embryogenesis and fertile green plant regeneration from suspension cell-derived protoplasts of rye (Secale cereale L. cv. Auvinen) was developed. Fast-growing and friable embryogenic calli with a high regeneration capacity were induced from immature rye inflorescences using
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service