Skip to Content
Merck
All Photos(1)

Documents

DNTP10

Sigma-Aldrich

Deoxynucleotide Set, 10 mM

Individual dNTPs for routine PCR; 0.5 mL each

Synonym(s):

10mM dNTP set, DNTP 10 mM solution, dNTP set, dNTPs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305
NACRES:
NA.52

form

liquid

concentration

10 mM

color

colorless

shipped in

dry ice

storage temp.

−20°C

General description

Deoxynucleoside triphosphates (dNTPs) are substrates for DNA polymerizing enzymes and are the essential building blocks of nucleic acid molecules. It aids in the synthesis of copies of the target DNA by providing each of the four bases (dATP, dGTP, dCTP, dTTP).(2) dNTP is a key component of PCR master mixes as it is necessary for DNA amplification reaction.

Application

Deoxynucleotide Set, 10 mM has been used in:
  • polymerase chain reaction (PCR)
  • nested PCR
  • DNA repair
  • the preparation of DNA-duplex

Packaging

0.5 mL each of 10 mM dATP, dCTP, dGTP and dTTP

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas A Gillet et al.
Methods in molecular biology (Clifton, N.J.), 1582, 127-141 (2017-03-31)
We describe here a method to identify the position of retroviral insertion sites and simultaneously to quantify the absolute abundance of each clone, i.e., the number of cells having the provirus inserted at a given place in the host genome.
Genetic (RAPD) diversity in two Armadillidium vulgare populations
Homor P, et al.
Tissue Antigens, 34, 19-22 (2003)
Thomas M Dechiara et al.
Methods in molecular biology (Clifton, N.J.), 530, 311-324 (2009-03-07)
With the completion of the human and mouse genome sequences and the development of high-throughput knockout mouse technologies, there is now a need for equally high-throughput methods for the production of mice for phenotypic studies. In response to this challenge
Chapter 8 - DNA Sequencing
Clark D P, et al.
Molecular Biology of the Gene, 240-269 (2019)
Jiannis Ragoussis et al.
Methods in molecular biology (Clifton, N.J.), 338, 261-280 (2006-08-05)
Recent studies on genome-wide localization of transcription factor (TF) binding to DNA have shown that a large proportion of identified sequences do not contain consensus motifs predicted by databases of transcription factor binding sites, such as TRANSFAC. The main limitation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service