Skip to Content
Merck

D6434

DMEM/F-12

With HEPES, sodium bicarbonate and sodium pyruvate, without ʟ-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME/Nutrient Mixture F-12 Ham, DMEM Hams F12, DME/F-12, 1:1 mixture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture

Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

L-glutamine: no
phenol red: no
NaHCO3: yes
HEPES: 15 mM
sodium pyruvate: 0.055 g/L
glucose: 3.15 g/L

shipped in

ambient

storage temp.

2-8°C

General description

Pyridoxal, which is an aldehyde, has been replaced with pyridoxine. This modification has been shown to improve the stability of the medium. Suitable for growth of a wide range of cell types in low serum conditions. Absence of phenol red makes this a preferred medium for clonal and stem cell culture.

Application

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham has been used to culture:

  • bovine endometrial epithelial cells
  • cancer stem cells
  • breast cancer cell line

Preparation Note

Supplement with 0.365 g/L L-glutamine


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Tânia C Gonçalves et al.
Neuropharmacology, 133, 404-414 (2018-02-24)
The Chinese bird spider huwentoxin-IV (HwTx-IV) is well-known to be a highly potent blocker of NaV1.7 subtype of voltage-gated sodium (NaV) channels, a genetically validated analgesic target, and thus promising as a potential lead molecule for the development of novel
Harshul Pandit et al.
BMC cancer, 18(1), 783-783 (2018-08-05)
Hepatocellular carcinoma (HCC) is among the deadliest cancers due to its heterogeneity, contributing to chemoresistance and recurrence. Cancer stem-like cells (CSCs) are suggested to play an important role in HCC tumorigenesis. This study investigates the role of Wnt/β-catenin pathway in
Racha Zgheib et al.
Cell death & disease, 10(8), 596-596 (2019-08-10)
Methionine dependency of tumor growth, although not well-understood, is detectable by 11C-methionine positron emission tomography and may contribute to the aggressivity of glioblastomas (GBM) and meningiomas. Cytosolic folate cycle is required for methionine synthesis. Its dysregulation may influence cell reprogramming



Global Trade Item Number

SKUGTIN
D6434-6X500ML04061833587591
D6434-500ML04061833587584