Skip to Content
Merck
All Photos(3)

Documents

D6277

Sigma-Aldrich

N-Decanoyl-N-methylglucamine

≥98% (GC)

Synonym(s):

N-(D-Glucityl)-N-methyldecanamide, N-Decanoyl-N-methyl-D-glucamine, MEGA-10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H35NO6
CAS Number:
Molecular Weight:
349.46
Beilstein:
6974359
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Assay

≥98% (GC)

form

powder

mol wt

349.46 g/mol

technique(s)

ligand binding assay: suitable
protein purification: suitable

CMC

6-7 mM (20-25°C)

storage temp.

2-8°C

SMILES string

OCC(O)C(O)C(O)C(O)CN(C)C(CCCCCCCCC)=O

InChI

1S/C17H35NO6/c1-3-4-5-6-7-8-9-10-15(22)18(2)11-13(20)16(23)17(24)14(21)12-19/h13-14,16-17,19-21,23-24H,3-12H2,1-2H3

InChI key

UMWKZHPREXJQGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Decanoyl-N-methylglucamine is a nonionic detergent used in lysis buffers.

Application

N-Decanoyl-N-methylglucamine has been used in a study to assess the effect of surfactants on wetting behavior of super-hydrophobic surfaces. It has also been used in a study to investigate the clinical impact of Human metapneumovirus (hMPV) vaccines on Syrian golden hamsters.
Non-ionic, dialyzable detergent used for the solubilization of membrane proteins

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sander Herfst et al.
The Journal of general virology, 88(Pt 10), 2702-2709 (2007-09-18)
Human metapneumovirus (hMPV), a newly discovered paramyxovirus, is associated with acute respiratory-tract illness, primarily in young children, individuals with underlying disease and the elderly. Two genetic lineages of hMPV circulate around the world, and viruses from these two lineages demonstrate
Khemraj Budachetri et al.
mBio, 11(4) (2020-07-30)
Ehrlichia chaffeensis is an obligatory intracellular bacterium that causes human monocytic ehrlichiosis, an emerging disease transmitted by the Lone Star tick, Amblyomma americanum. E. chaffeensis outer membrane protein entry triggering protein of Ehrlichia (EtpE) is necessary for bacterial entry into
R Mohammadi et al.
Langmuir : the ACS journal of surfaces and colloids, 20(22), 9657-9662 (2004-10-20)
The effect of surfactants on wetting behavior of super-hydrophobic surfaces was investigated. Super-hydrophobic surfaces were prepared of alkylketene dimer (AKD) by casting the AKD melt in a specially designed mold. Time-dependent studies were carried out, using the axisymmetric drop shape
R C Poole et al.
The Biochemical journal, 283 ( Pt 3), 855-862 (1992-05-01)
1. Intact erythrocytes were incubated with 100 microM-4,4'-di-isothiocyanostilbene-2,2'-disulphonate (DIDS), a concentration sufficient to inhibit lactate transport irreversibly by 65%. DIDS-labelled proteins were detected by immunoblotting of erythrocyte membrane proteins with an anti-DIDS antibody. Labelled polypeptides of 35-45 kDa in rat
C Carnero Ruiz et al.
Biomacromolecules, 8(8), 2497-2503 (2007-07-17)
The protein-surfactant system constituted by bovine serum albumin (BSA) and N-decanoyl-N-methylglucamide (MEGA-10) has been studied by using surface tension, steady-state fluorescence, and dynamic light scattering measurements. It was found that the presence of protein delays the surfactant aggregation, which was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service