Skip to Content
Merck
All Photos(2)

Key Documents

A7154

Sigma-Aldrich

Adenosine, periodate oxidized

≥93%

Synonym(s):

ADOX, Adenosine-2′,3′-dialdehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5O4
CAS Number:
Molecular Weight:
265.23
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥93%

form

powder

solubility

0.2 M HCl: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)C(OC(CO)C=O)C=O

InChI

1S/C10H11N5O4/c11-9-8-10(13-4-12-9)15(5-14-8)7(3-18)19-6(1-16)2-17/h1,3-7,17H,2H2,(H2,11,12,13)

InChI key

ILMNSCQOSGKTNZ-UHFFFAOYSA-N

Application

Adenosine, periodate oxidized has been used:
  • as a methylarginine transferase inhibitor in the human embryonic kidney (HEK)-293 T cells
  • as a methylase inhibitor in H4 neuroglioma
  • as a broad inhibitor of S-adenosylmethionine (AdoMet)-dependent methyltransferases in mouse embryo fibroblast NIH3T3 cells

Biochem/physiol Actions

Adenosine, periodate oxidized (Adox) is a protein arginine methyltransferases (PRMTs) inhibitor. It also inhibits the enzyme S-adenosylhomocysteine hydrolase and induces apoptosis. Its inhibitory effect on histone methyltransferases prevents histone methylation. Adox also elicits intrinsic cytotoxic properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinghong He et al.
Journal of translational medicine, 11, 14-14 (2013-01-16)
Pharmacologic reactivation of fetal hemoglobin expression is a promising strategy for treatment of sickle cell disease and β-thalassemia. The objective of this study was to investigate the effect of the methyl transferase inhibitor adenosine-2',3'-dialdehyde (Adox) on induction of human fetal
Jin-Ah Park et al.
Molecules and cells, 31(4), 343-349 (2011-03-02)
Interphasic chromatin condenses into the chromosomes in order to facilitate the correct segregation of genetic information. It has been previously reported that the phosphorylation and methylation of the N-terminal tail of histone H3 are responsible for chromosome condensation. In this
Ryan Heit et al.
Journal of cell science, 122(Pt 16), 2957-2968 (2009-07-30)
Trimethylation of lysine 9 on histone H3 (H3K9me3) is known both to be necessary for proper chromosome segregation and to increase in late G2. We investigated the role of late G2 methylation, specifically in mitotic progression, by inhibiting methylation for
Alla Polotskaia et al.
Cell cycle (Georgetown, Tex.), 6(20), 2524-2530 (2007-08-30)
With the recent characterization of enzymes responsible for protein arginine methylation and demonstration that catabolic products of arginine methylation, such as asymmetric dimethylarginine (ADMA), are among the most powerful mechanisms of atherogenesis, developing endothelial dysfunction and cardiovascular complications in a
Christian Schwerk et al.
Oncogene, 24(47), 7002-7011 (2005-07-12)
We have analysed the importance of proper substrate methylation by S-adenosylmethionine-dependent methyltransferases for cell survival and cell cycle progression. We show that treatment of cells with the methyltransferase inhibitor adenosine dialdehyde (AdOx) causes cell cycle arrest and death in different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service