Skip to Content
Merck
All Photos(2)

Key Documents

D3385

Sigma-Aldrich

3′-Dephosphocoenzyme A

≥90% (HPLC)

Synonym(s):

3′-Desphosphocoenzyme A, dpCoA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H35N7O13P2S
CAS Number:
Molecular Weight:
687.55
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥90% (HPLC)

form

powder

impurities

≤1% CoA

solubility

H2O: soluble-50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(C)(COP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1O)n2cnc3c(N)ncnc23)C(O)C(=O)NCCC(=O)NCCS

InChI

1S/C21H35N7O13P2S/c1-21(2,16(32)19(33)24-4-3-12(29)23-5-6-44)8-39-43(36,37)41-42(34,35)38-7-11-14(30)15(31)20(40-11)28-10-27-13-17(22)25-9-26-18(13)28/h9-11,14-16,20,30-32,44H,3-8H2,1-2H3,(H,23,29)(H,24,33)(H,34,35)(H,36,37)(H2,22,25,26)

InChI key

KDTSHFARGAKYJN-UHFFFAOYSA-N

Application

3′-Dephosphocoenzyme A has been used as a substrate to initiate transcription.
Dephosphocoenzyme A is the direct precursor of coenzyme A. It is used for the synthesis of coenzyme A by bifunctional CoA Synthase (CoASy, 4′-phosphopantetheine adenylyltransferase/dephospho-CoA kinase) which performs the transfer of the γ-phosphate of ATP to dephosphocoenzyme A.

Biochem/physiol Actions

3′-Dephosphocoenzyme A (DepCoA), also called desphosphoCoA (dpCoA), is synthesized from 4-phosphopantetheine by the enzyme phosphopanthetheine adenylyltransferase. It is an intermediate in coenzyme A biosynthetic pathway. In the presence of dephospho-CoA kinase (CoaE), dephosphocoenzyme A is converted to coenzyme A. 3′-Dephosphocoenzyme shares adenosine-diphosphate substructure as like the purines and is a non-canonical nucleotide and is an initiator of transcription initiation. It is routinely used in in vitro transcription studies to initiate RNA synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mechanism of RNA 5? capping with NAD+, NADH and desphospho-CoA
Bird JG, et al.
Nature, 535(7612), 444-444 (2016)
A Geerlof et al.
The Journal of biological chemistry, 274(38), 27105-27111 (1999-09-10)
Phosphopantetheine adenylyltransferase (PPAT) catalyzes the penultimate step in coenzyme A (CoA) biosynthesis: the reversible adenylation of 4'-phosphopantetheine yielding 3'-dephospho-CoA and pyrophosphate. Wild-type PPAT from Escherichia coli was purified to homogeneity. N-terminal sequence analysis revealed that the enzyme is encoded by
Thomas J Wubben et al.
Journal of molecular biology, 404(2), 202-219 (2010-09-21)
Phosphopantetheine adenylyltransferase (PPAT) catalyzes the penultimate step in the coenzyme A (CoA) biosynthetic pathway, reversibly transferring an adenylyl group from ATP to 4'-phosphopantetheine (PhP) to form dephosphocoenzyme A. This reaction sits at the branch point between the de novo pathway
Azusa Seto et al.
Proteins, 58(1), 235-242 (2004-11-05)
Dephosphocoenzyme A kinase (DCK) catalyzes phosphorylation in the final step of coenzyme A (CoA) biosynthesis. In this phosphorylation process, domain movements play a very important role. To reveal the structural changes induced by ligand binding, we determined the crystal structure
Efficient one-pot enzymatic synthesis of dephospho coenzyme A
Sapkota K and Huang F
Bioorganic Chemistry, 76, 23-27 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service