Skip to Content
Merck
All Photos(2)

Key Documents

D2915

Sigma-Aldrich

Dexamethasone-Water Soluble

suitable for cell culture, BioReagent

Synonym(s):

Dexamethasone – Cyclodextrin complex

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic (organic)
form:
solid

biological source

synthetic (organic)

product line

BioReagent

form

solid

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 25 mg/mL, clear to slightly hazy

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@@H]1CC2C3CCC4=CC(=O)C=C[C@]4(C)C3[C@@H](O)C[C@]2(C)[C@H]1C(=O)CO.OC[C@H]5O[C@@H]6O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]9[C@H](O)[C@@H](O)[C@H](O[C@@H]9CO)O[C@H]%10[C@H](O)[C@@H](O)[C@H](O[C@@H]%10CO)O[C@H]%11[C@H](O)[C@@H](O)[C@H](O[C@@H]%11CO)O[C@H]%12[C@H](O)[C@@H](O)[C@H](O[C@@H]%12CO)O[C@H]5[C@H](O)[C@H]6O

InChI

1S/C42H70O35.C22H30O4/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51;1-12-8-16-15-5-4-13-9-14(24)6-7-21(13,2)20(15)17(25)10-22(16,3)19(12)18(26)11-23/h8-63H,1-7H2;6-7,9,12,15-17,19-20,23,25H,4-5,8,10-11H2,1-3H3/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-;12-,15+,16?,17+,19-,20-,21+,22+/m11/s1

InChI key

XEJNDGTUKUMQNG-ARJMFYCQSA-N

Application

Dexamethasone-Water Soluble has been used as a component of
  • Dulbecco′s modified Eagle medium based osteogenic and adipogenic medium for human mesenchymal stem cell culture
  • erythroid expansion media for the growth of liver, bone marrow and spleen tissues
  • medium 199 (M199) for hepatocyte culture

Biochem/physiol Actions

Dexamethasone is an anti-inflammatory glucocorticoid and a synthetic corticosteroid used for treating corneal inflammation. It inhibits the IL-1β-dependent angiogenesis. Dexamethasone elicits a range of effects on cell survival, cell signaling and gene expression. It favors apoptosis in chondrocytes and impairs development of bone. Dexamethasone is a potential therapeutic agent as it induces apoptosis in multiple myeloma (MM) cells. It elicits its functionality by lowering glucocorticoid receptor (GR) α levels and inhibits colon tumor progression.

Packaging

Package size based on dexamethasone

Other Notes

Contains approx. 65 mg dexamethasone per gram; balance methyl-β-cyclodextrin.

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of temperature on the viability of human mesenchymal stem cells
Reissis Y, et al.
Stem Cell Research & Therapy, 4(6), 139-139 (2013)
Mechanisms of dexamethasone-mediated inhibition of Toll-like receptor signaling induced by Neisseria meningitidis and Streptococcus pneumoniae
Mogensen TH, et al.
Infection and Immunity, 76(1), 189-197 (2008)
Tissue-specific actions of glucocorticoids on apoptosis: a double-edged sword
Gruver-Yates A and Cidlowski J
Cells, 2(2), 202-223 (2013)
Immature erythroblasts with extensive ex vivo self-renewal capacity emerge from the early mammalian fetus
England SJ, et al.
Blood, 117(9), 2708-2717 (2011)
Plasma membrane phospholipase A2 controls hepatocellular fatty acid uptake and is responsive to pharmacological modulation: implications for nonalcoholic steatohepatitis
Stremmel W, et al.
Faseb Journal, 28(7), 3159-3170 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service