Skip to Content
Merck
All Photos(1)

Documents

D1408

Sigma-Aldrich

Dulbecco′s Phosphate Buffered Saline

Liquid (10X), without MgCl₂ and CaCl₂, sterile-filtered, suitable for cell culture

Synonym(s):

DPBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
eCl@ss:
32129211
NACRES:
NA.75

product name

Dulbecco′s Phosphate Buffered Saline, 10×, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

sodium pyruvate: no
glucose: no (Dextro)

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Dulbecco′s Phosphate Buffered Saline (DPBS) is a balanced salt solution.

Application

Dulbecco′s Phosphate Buffered Saline has been used:
  • in microfluidic cell culture of mouse embryonic stem cells (mESC)
  • to dissolve albumin for adsorption studies
  • in PC12 cell culture experiments

Dulbecco′s Phosphate Buffered Saline 10X is a stock solution used to prepare 1X D-PBS in cell culture grade water (W3500). D-PBS is used to wash cells during preparation and serial transfer.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah M Hicks et al.
The Journal of infectious diseases, 223(1), 10-14 (2020-10-04)
Estimates of seroprevalence of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) antibodies have been hampered by inadequate assay sensitivity and specificity. Using an enzyme-linked immunosorbent assay-based approach that combines data about immunoglobulin G responses to both the nucleocapsid and spike
Alexandre Super et al.
Biotechnology journal, 11(9), 1179-1189 (2016-05-24)
Oxygen plays a key role in stem cell biology as a signaling molecule and as an indicator of cell energy metabolism. Quantification of cellular oxygen kinetics, i.e. the determination of specific oxygen uptake rates (sOURs), is routinely used to understand
Marjan Nabili et al.
Ultrasound in medicine & biology, 39(4), 638-646 (2013-02-19)
Delivery of sufficient amounts of therapeutic drugs into the eye is often a challenging task. In this study, ultrasound application (frequencies of 400 KHz to 1 MHz, intensities of 0.3-1.0 W/cm(2) and exposure duration of 5 min) was investigated to overcome
A D Cabral et al.
The Analyst, 142(20), 3922-3933 (2017-09-21)
Chemosensors for the detection of phosphate-containing biological species are in high need. Detection of proximally phosphorylated sites of PP
Yuta Tanaka et al.
Journal of biomedical materials research. Part A, 92(1), 350-358 (2009-02-04)
Both terminals of a poly(ethylene glycol) (PEG) molecule were terminated with amines. The PEG was electrodeposited onto titanium (Ti) to give it the biofunctions such as blood compatibility. The effects of the amine of PEG terminals and the pH at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service