Skip to Content
Merck
All Photos(1)

Key Documents

P5493

Sigma-Aldrich

Phosphate buffered saline

10X, solution, pH 7.2-7.6 (25 °C), BioPerformance Certified, for molecular biology

Synonym(s):

PBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
NACRES:
NA.25

Product Name

Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture

grade

BioPerformance Certified
Molecular Biology
for molecular biology

Quality Level

100
200

sterility

0.2 μm filtered

form

solution

storage condition

(Tightly closed)

technique(s)

cell culture | mammalian: suitable
immunocytochemistry: suitable
immunofluorescence: suitable

impurities

Bioburden, tested
endotoxin, tested
≤5 ppm heavy metals (as Pb)

color

colorless

pH

7.2-7.6 (25 °C)

solubility

water: soluble

cation traces

heavy metals (as Pb): ≤5 ppm

suitability

suitable for
suitable for flow cytometry
suitable for microscopy

application(s)

advanced drug delivery

foreign activity

DNAse, none detected
NICKase, none detected
Protease, none detected
RNAse, none detected

SMILES string

OP(=O)(O)[O-].OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Cl-].[Cl-].[K+].[K+]

InChI

InChI=1S/2ClH.2K.3Na.2H3O4P/c;;;;;;;2*1-5(2,3)4/h2*1H;;;;;;2*(H3,1,2,3,4)/q;;5*+1;;/p-5

InChI key

LOKCTEFSRHRXRJ-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Application

Phosphate buffered saline has been used:
  • as a cell wash solution in human dental pulp stem cells (DPSCs)
  • for perfusion studies in dissected brain samples
  • as wash solution for neurons culture cells sample preparation for immunofluorescence and immunocytochemical labelling
  • as wash solution for ovary and sperm during in vitro culture and fertilization

Packaging

4L package size comes in a box with a spigot for spill free dispensing.

Other Notes

When diluted to a 1X concentration, this product will yield a phosphate buffered saline solution with a phosphate buffer concentration of 0.01M and a sodium chloride concentration of 0.154M. The solution pH will be 7.4.
For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differences in serum steroid hormones concentrations in spontaneously hypertensive
Kozlowska A, et al.
Physiological Research (2018)
Maturation, Fertilization and Implantation of Cattle Ova Cultured in Different Media
Saif AH, et al.
The eurasia proceedings of science, technology, engineering & mathematics., 3(9), 181-188 (2018)
Diverse effect of BMP-2 homodimer on mesenchymal progenitors of different origin
Hrubi E, et al.
Human Cell : Official Journal of Human Cell Research Society, 31(2), 139-148 (2018)
Stephen J Bush et al.
G3 (Bethesda, Md.), 9(2), 359-373 (2018-12-12)
One of the most significant physiological challenges to neonatal and juvenile ruminants is the development and establishment of the rumen. Using a subset of RNA-Seq data from our high-resolution atlas of gene expression in sheep (Ovis aries) we have provided
Distribution and Neurochemistry of the Porcine Ileocaecal Valve Projecting Sensory Neurons in the Dorsal Root Ganglia and the Influence of Lipopolysaccharide from Different Serotypes of Salmonella spp. on the Chemical Coding of DRG Neurons in the Cell Cultures
Mikolajczyk A, et al.
International Journal of Molecular Sciences, 19(9), 2551-2551 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service