CRM46911
Deoxynivalenol solution
certified reference material, 200 μg/mL in ethyl acetate: methanol (95:5), ampule of 1 mL
Synonym(s):
Deoxynivalenol solution, DON, DON solution, 3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, Vomitoxin
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
CofA
current certificate can be downloaded
packaging
ampule of 1 mL
concentration
200 μg/mL in ethyl acetate: methanol (95:5)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
single component solution
storage temp.
-10 to -25°C
SMILES string
CC1=C[C@H]2O[C@@H]3[C@H](O)C[C@@](C)([C@]34CO4)[C@@]2(CO)[C@H](O)C1=O
InChI
1S/C15H20O6/c1-7-3-9-14(5-16,11(19)10(7)18)13(2)4-8(17)12(21-9)15(13)6-20-15/h3,8-9,11-12,16-17,19H,4-6H2,1-2H3/t8-,9-,11-,12-,13-,14-,15+/m1/s1
InChI key
LINOMUASTDIRTM-QGRHZQQGSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Deoxynivalenol is a naturally occurring type B trichothecene that belongs to the class of Fusarium toxins. Mostly found in cereal grains, it is a sesquiterpenoid polar compound and has a carbonyl group at C-8.
Check out our entire range of trichothecene type mycotoxin reference materials
Application
- Determination of total deoxynivalenol in human urine samples of 203 volunteers, consisting of six population groups, by ultra high-performance liquid chromatography coupled with mass spectrometric detection (UHPLC-MS)
- Quantification of deoxynivalenol and aflatoxin M1 in human urine samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) and high-performance liquid chromatography-fluorescence detection (HPLC-FD) respectively
- Simultaneous estimation of two trichothecenes— deoxynivalenol and T-2 toxin, in 11 maize and 10 oats samples by UHPLC combined with triple quadrupole mass spectrometer
- Quantitative analysis of deoxynivalenol and deepoxy-deoxynivalenol in pig colostrum and serum samples by solid phase extraction combined with UHPLC-high resolution tandem mass spectrometry (HRMS/MS)
- Immunoaffinity column-based cleanup and HPLC-UV method-based determination of nivalenol and deoxynivalenol in white rice and brown rice samples
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2 - STOT SE 3
Target Organs
Central nervous system, Eyes
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
26.6 °F - closed cup
Flash Point(C)
-3 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service