Skip to Content
Merck
All Photos(2)

Key Documents

C9658

Sigma-Aldrich

Carboxypeptidase G from Pseudomonas sp.

lyophilized powder, ≥3 units/mg protein

Synonym(s):

γ-Glutamyl hydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥3 units/mg protein

mol wt

homodimer ~90 kDa

composition

Protein, ~70% biuret

solubility

H2O: soluble 0.5 mg/mL

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Carboxypeptidase G from Pseudomonas sp., or γ-Glutamyl hydrolase, has been used in a study to assess the role of the putidaredoxin COOH-terminus in P-450cam (cytochrome m) hydroxylations. Carboxypeptidase G from Pseudomonas sp. has also been used in a study to investigate the effects of nitric oxide on pemetrexed cytotoxicity via NO‑cGMP signaling in lung adenocarcinoma cells.

Biochem/physiol Actions

Carboxypeptidase G is a lysosomal, thiol-dependent protease, which progressively cleaves γ-glutamyl pteroyl poly-γ-glutamate yielding pteroyl-α-glutamate (folic acid) and free glutamate. It is considered highly specific for the γ-glutamyl bond, but not for the C-terminal amino acid of the leaving group. Molecular mass of this homodimer is approximately 90 kDa. The enzyme is activated by Zn2+ ions.

Unit Definition

One unit will hydrolyze 1.0 μmole of L-glutamic acid from (+)amethopterin per min at pH 7.3 at 30 °C.

Physical form

Contains sodium acetate salt.

Preparation Note

Chromatographically purified
Solutions should be prepared fresh prior to use.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edgar M Pera et al.
Gene expression patterns : GEP, 3(2), 147-152 (2003-04-25)
Endoderm development is an area of intense interest in developmental biology, but progress has been hampered by the lack of specific markers for differentiated endodermal cells. In an unbiased secretion cloning screen of Xenopus gastrula embryos we isolated a novel
The enzymatic hydrolysis of methotrexate and folic acid.
C C Levy et al.
The Journal of biological chemistry, 242(12), 2933-2938 (1967-06-25)
Jeremy P Mallari et al.
Bioorganic & medicinal chemistry, 12(22), 6011-6020 (2004-10-23)
A series of alkyl and aryl phosphonyl, thiophosphonyl, and dithiophosphonyl derivatives of (S)- and (R)-glutamic acid were prepared and examined for inhibitory potency against glutamate carboxypeptidase (carboxypeptidase G). The acquisition of the phosphonamidodithioic acids and the individual phosphonamidothioic acid diastereomers
Takashi Tsukamoto et al.
Bioorganic & medicinal chemistry letters, 12(16), 2189-2192 (2002-07-20)
Phosphonate and phosphinate analogues of N-acylated gamma-glutamylglutamate were tested for the ability to inhibit glutamate carboxypeptidase II (GCP II). All of the compounds inhibit GCP II with IC(50) values in the low nanomolar range. The comparison of the results to
Masaharu Suzuki et al.
Current opinion in plant biology, 11(5), 548-553 (2008-08-12)
Two subfamilies of plant-specific B3 domain transcription factors regulate the fundamental transition between seed and vegetative phases of development. The AFL B3 genes activate the embryo maturation program, while the closely related VAL B3 genes shutdown the AFL network before

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service