Skip to Content
Merck
All Photos(1)

Documents

C9630

Sigma-Aldrich

Cytidine 2′:3′-cyclic monophosphate monosodium salt

≥95% (HPLC)

Synonym(s):

2′,3′-cCMP, 2′,3′-Cyclic CMP monosodium salt, Cyclic cytidylic acid, Cytidine 2′,3′-cyclophosphate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N3NaO7P
CAS Number:
Molecular Weight:
327.16
Beilstein:
4086446
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic

Assay

≥95% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@H]3OP([O-])(=O)O[C@@H]23

InChI

1S/C9H12N3O7P.Na/c10-5-1-2-12(9(14)11-5)8-7-6(4(3-13)17-8)18-20(15,16)19-7;/h1-2,4,6-8,13H,3H2,(H,15,16)(H2,10,11,14);/q;+1/p-1/t4-,6-,7-,8-;/m1./s1

InChI key

SQOIXCJUYWSZDW-IAIGYFSYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cytidine 2′,3′-cyclic monophosphate (2′,3′-CyclicCMP) (2′,3′-cCMP) is used as a model substrate for kinetic analysis of various ribonucleases, especially ribonuclease A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shawn D Spencer et al.
Pharmaceutical research, 21(9), 1642-1647 (2004-10-23)
Isothermal titration calorimetry (ITC) and progress curve analysis was used to measure the enzyme kinetic parameters (KM and kcat) of the hydrolysis of cCMP by RNase-A, a reaction that includes end-product competitive inhibition by 3'-CMP. The heat generated from injection
M Ribó et al.
Protein expression and purification, 7(3), 253-261 (1996-05-01)
Human pancreatic ribonuclease (HP-RNase) has considerable promise as a therapeutic agent. Structure-function analyses of HP-RNase have been impeded by the difficulty of obtaining the enzyme from its host. Here, a gene encoding HP-RNase was designed, synthesized, and inserted into two
Y Markert et al.
Protein engineering, 14(10), 791-796 (2001-12-12)
Although highly stable toward unfolding, native ribonuclease A is known to be cleaved by unspecific proteases in the flexible loop region near Ala20. With the aim to create a protease-resistant ribonuclease A, Ala20 was substituted for Pro by site-directed mutagenesis.
S Adinolfi et al.
FEBS letters, 398(2-3), 326-332 (1996-12-02)
In the ribonuclease superfamily, dimericity is a unique feature of bovine seminal RNase (BS-RNase). In about two-thirds of native BS-RNase molecules, the two subunits interchange their N-terminal tails, thus generating domain-swapped dimers (MxM), which mostly responsible for enzyme biological activities
Mojca Bencina et al.
Journal of chromatography. A, 1144(1), 135-142 (2007-01-20)
In gene therapy and DNA vaccination, RNA removal from DNA preparations is vital and is typically achieved by the addition of ribonuclease into the sample. Removal of ribonuclease from DNA samples requires an additional purification step. An alternative is the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service