Skip to Content
Merck
All Photos(3)

Key Documents

C8128

Sigma-Aldrich

5α-Cholestan-3-one

crystalline

Synonym(s):

3-Keto-5α-cholestane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

crystalline

Quality Level

mp

128-130 °C (lit.)

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCCC(C)C)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CCC(=O)C2

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-20,22-25H,6-17H2,1-5H3/t19-,20+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

PESKGJQREUXSRR-UXIWKSIVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dina S El-Kady et al.
Applied biochemistry and biotechnology, 188(3), 635-662 (2019-01-08)
Modification of steroid molecules by introducing heterocyclic ring into the core structure of steroids has been utilized as an attractive approach for either cancer prognosis or diagnosis. Several new cholestanoheterocyclic steroids were synthesized, and analytical and spectral data proved the
P P Nair et al.
The American journal of clinical nutrition, 40(4 Suppl), 931-936 (1984-10-01)
Cholesterol and its metabolites, together with bile acids, are implicated as risk factors in the genesis and progression of colon cancer. This study was designed to determine differences in the neutral sterol composition of stools from four different population groups
A K Bhattacharyya et al.
Nutrition and cancer, 12(1), 69-73 (1989-01-01)
This paper reports the differences in the fecal excretion of cholesterol and its degradation products in high- and low-responding rhesus monkeys fed diets with and without extra cholesterol. The high-responding monkeys had a great increase in plasma cholesterol concentration when
W F Lau et al.
Experientia, 51(7), 731-737 (1995-07-14)
The effects of cholesterol and cholesterol-derived oxysterols (cholestanone, cholestenone, coprostanone and epicoprostanol) on adipocyte ghost membrane fluidity were studied using a fluorescence depolarization method. The fluorescence anisotropy of the treated membranes was determined using 1,6-diphenyl-1,3,5-hexatriene (DPH) and 1-(4-trimethylammoniumphenyl)-6-phenyl-1,3,5-hexatriene (TMA-DPH). Cholestanone
Bianka Ditscheid et al.
European journal of nutrition, 48(1), 22-30 (2008-11-15)
Previous human studies on the effect of dietary calcium supplementation on faecal excretion of bile acids (BA) and faecal water concentrations of animal neutral sterols (NSt, cholesterol and its metabolites) lack detailed information about single BA and NSt. We investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service