Skip to Content
Merck
All Photos(1)

Key Documents

188174

Sigma-Aldrich

(+)-4-Cholesten-3-one

98%

Synonym(s):

3-Keto-4-cholestene, 3-Oxo-4-cholestene, 4-Cholesten-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
Beilstein:
2224119
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]23/D +91.0°, c = 2 in chloroform

mp

79-81 °C (lit.)

functional group

ketone

SMILES string

[H][C@@]12[C@]([C@](CC3)(C)C(CC2)=CC3=O)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC(C)C

InChI

1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h17-19,22-25H,6-16H2,1-5H3/t19-,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

NYOXRYYXRWJDKP-GYKMGIIDSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daniel L Motola et al.
Cell, 124(6), 1209-1223 (2006-03-15)
In response to environmental and dietary cues, the C. elegans orphan nuclear receptor, DAF-12, regulates dauer diapause, reproductive development, fat metabolism, and life span. Despite strong evidence for hormonal control, the identification of the DAF-12 ligand has remained elusive. In
G Tibolla et al.
Atherosclerosis, 210(1), 78-87 (2009-12-01)
Atherosclerosis is associated with Alzheimer's disease (AD) in humans, but the nature of this link is still elusive. Aim of this study was to investigate aortic atherosclerosis development in a mouse model with central nervous system (CNS) restricted beta-amyloid precursor
Yin-Ru Chiang et al.
Applied and environmental microbiology, 74(1), 107-113 (2007-11-13)
The anoxic metabolism of cholesterol was studied in the denitrifying bacterium Sterolibacterium denitrificans, which was grown with cholesterol and nitrate. Cholest-4-en-3-one was identified before as the product of cholesterol dehydrogenase/isomerase, the first enzyme of the pathway. The postulated second enzyme
Satoshi Yamaguchi et al.
Scientific reports, 7, 41007-41007 (2017-01-25)
A chemically-activatable alkynyl steroid analogue probe has been synthesized for visualizing the lipid raft membrane domains by Raman microscopy. The Raman probe, in which ring A of its steroid backbone is replaced with an alkynyl group, was designed to enable
Liang-Bin Xiong et al.
Microbial cell factories, 19(1), 80-80 (2020-04-02)
The bioconversion of phytosterols into high value-added steroidal intermediates, including the 9α-hydroxy-4-androstene-3,17-dione (9-OHAD) and 22-hydroxy-23,24-bisnorchol-4-ene-3-one (4-HBC), is the cornerstone in steroid pharmaceutical industry. However, the low transportation efficiency of hydrophobic substrates into mycobacterial cells severely limits the transformation. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service