Skip to Content
Merck
All Photos(2)

Key Documents

A2480

Sigma-Aldrich

5α-Androstan-3β-ol

powder

Synonym(s):

3β-Hydroxy-5α-androstane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H32O
CAS Number:
Molecular Weight:
276.46
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

form

powder

solubility

DMSO: soluble 14 mg/mL at ≤60 °C
H2O: insoluble

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CCC[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@H](O)C2

InChI

1S/C19H32O/c1-18-9-3-4-16(18)15-6-5-13-12-14(20)7-11-19(13,2)17(15)8-10-18/h13-17,20H,3-12H2,1-2H3/t13-,14-,15-,16-,17-,18-,19-/m0/s1

InChI key

DJTOLSNIKJIDFF-LOVVWNRFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5α-Androstan-3β-ol has been used as a cortisone analog to test its effect on the voltage-dependent potassium channel (Kv) current.

Biochem/physiol Actions

mCAR (constitutive androstane receptor) inverse agonist; testosterone metabolite.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D B Gower et al.
The Journal of steroid biochemistry and molecular biology, 48(4), 409-418 (1994-03-01)
The concentrations of five 16-androstene steroids were determined, by a GC-MS method, in freshly-produced apocrine sweat (adrenaline-induced), in 8 men and 2 women. The ranges of concentrations (nmol/microliter) in apocrine sweat were: 5 alpha-androst-16-en-3-one (5 alpha-A), 0.1-2.0 and 4,16-androstadien-3-one (androstadienone)
Yaping Pan et al.
ACS chemical biology, 7(10), 1641-1646 (2012-07-19)
The Kv1 family voltage-dependent K(+) channels are essential for termination of action potentials in neurons and myocytes. These channels form a stable complex with their beta subunits (Kvβ), some of which inhibit channel activity. Cortisone potentiates Kv1 channel by binding
Andrea Toell et al.
Journal of cellular biochemistry, 85(1), 72-82 (2002-03-14)
Constitutive androstane receptor (CAR) and pregnane X receptor (PXR) are members of the nuclear receptor superfamily that regulate target gene transcription in a ligand-dependent manner. CAR and PXR have a rather broad, overlapping set of ligands that range from natural
Giovanna M Ledda-Columbano et al.
Carcinogenesis, 24(6), 1059-1065 (2003-06-17)
The nuclear receptor Constitutive Androstane Receptor (CAR) binds DNA as a heterodimer with the retinoic-X receptor and activates gene transcription. Previously, in vitro studies have shown that the testosterone metabolites, androstenol and androstenol, inhibit the constitutive transcriptional activity of CAR
Ping Li et al.
British journal of pharmacology, 158(5), 1322-1329 (2009-08-26)
Potentiating neurosteroids are some of the most efficacious modulators of the mammalian GABA(A) receptor. One of the crucial interactions may be between the C20 ketone group (D-ring substituent at C17) of the neurosteroid, and the N407 and Y410 residues in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service