Skip to Content
Merck

Skip To

75199

Azithromycin

broad-spectrum antimicrobial agent, powder

Synonym(s):

CP 62,993

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C38H72N2O12
CAS Number:
Molecular Weight:
748.98
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Azithromycin,

form

powder

Quality Segment

optical activity

[α]/D −45±2°, c = 1% in ethanol

antibiotic activity spectrum

viruses

application(s)

environmental

mode of action

protein synthesis | interferes

SMILES string

CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C38H72N2O12/c1-15-27-38(10,46)31(42)24(6)40(13)19-20(2)17-36(8,45)33(52-35-29(41)26(39(11)12)16-21(3)48-35)22(4)30(23(5)34(44)50-27)51-28-18-37(9,47-14)32(43)25(7)49-28/h20-33,35,41-43,45-46H,15-19H2,1-14H3/t20-,21-,22+,23-,24-,25+,26+,27-,28+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1

InChI key

MQTOSJVFKKJCRP-BICOPXKESA-N

General description

Azithromycin is a hydrophobic azalide and a derivative of erythromycin. It belongs to the macrolide class of antibodies.

Application

Azithromycin has been used:

  • to treat synchronized Plasmodium falciparum Dd2 parasites for continuous growth assays to study the antiparasitic mechanisms of doxycycline (DOX)
  • to study its bactericidal activity against Pseudomonas aeruginosa in combination with a recombinant hemolymph plasma lectin (rHPLOE)
  • in evolution experiments to study P. aeruginosa strain PA14’s response to bacteriostatic antibiotics

Biochem/physiol Actions

Azithromycin is a broad-spectrum antimicrobial agent, effective against infections associated with respiratory tract, skin, tissues, and genital chlamydia. It acts against Gram-negative bacteria (including Enterobacteriaceae) and several Gram-positive organisms. It also acts against Plasmodium falciparum and P. vivax, which are multidrug resistance species that transmit malaria. Azithromycin targets the plasmodial ribosome and prevents protein synthesis. It is soluble in lipids and metabolizes in the liver by undergoing demethylation. It is commonly used to treat community-acquired pneumonia, sexually transmitted diseases, acute otitis media, and acute exacerbation of chronic obstructive pulmonary disease (COPD).

Features and Benefits

  • Rapidly absorbed
  • Shows great tissue penetration and procures high intracellular concentration
  • Distributes widely in body fluids and tissues

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1046056Y0000306PHR1088
form

powder

form

-

form

-

form

-

Quality Level

200

Quality Level

-

Quality Level

-

Quality Level

300

antibiotic activity spectrum

viruses

antibiotic activity spectrum

-

antibiotic activity spectrum

-

antibiotic activity spectrum

-

optical activity

[α]/D −45±2°, c = 1% in ethanol

optical activity

-

optical activity

-

optical activity

-

mode of action

protein synthesis | interferes

mode of action

-

mode of action

-

mode of action

-

application(s)

environmental

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)


pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Chronic 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the potency of Azithromycin 75199?

    1 answer
    1. Unfortunately, the potency of this product is not determined. The minimum purity specification for this compound is 95%.

      Helpful?

Reviews

No rating value

Active Filters