Skip to Content
Merck

Skip To

C7602

L-Cystine

98.5-101.0%, suitable for cell culture, BioXtra, non-animal source

Synonym(s):

(R,R)-3,3′-Dithiobis(2-aminopropionic acid)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30
NACRES:
NA.26
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
200-296-3
MDL number:
Beilstein/REAXYS Number:
1728094
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%

biological source

non-animal source

Quality Level

agency

meets EP testing specifications

product line

BioXtra

assay

98.5-101.0%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white to off-white

mp

>240 °C (dec.) (lit.)

solubility

1 M HCl: 100 mg/mL

application(s)

peptide synthesis

SMILES string

N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

LEVWYRKDKASIDU-IMJSIDKUSA-N

Application

L-Cystine has been used as a supplement in astroglial-conditioned media to culture cerebral astroglia. It has been used in several techniques, such as HPLC-FD, UHPLC-UV, UHPLC-MS, and triple quadrupole tandem mass spectrometry (UHPLC-MS/MS), to differentiate BMAA (β-N-methylamino-L-alanine), the cyanobacterial neurotoxin, from other diamino acids.
L-Cystine has been used as a supplement in oocyte culture media to improve oocyte maturation and parthenogenesis of embryonic development.

Biochem/physiol Actions

Cystinuria is characterized with renal transport and excretion of L-Cystine. Cysteine is one of the functional amino acids that are associated with growth, reproduction, maintenance and immunity. Cysteine is a source of disulfide linkage in protein and is associated with sulfur transport. At physiological pH, cysteine undergoes rapid oxidation to form cystine. Reduced availability of cysteine or cystine, influences leukocyte metabolism. L-Cystine is necessary for oxygen production and LDL (low density lipoprotein) modification by arterial smooth muscle cells.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C875530200779539
form

powder

form

crystalline

form

powder or crystals

form

powder

assay

98.5-101.0%

assay

≥98% (TLC)

assay

≥99.7% (TLC)

assay

≥98.5%

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

technique(s)

-

product line

BioXtra

product line

-

product line

-

product line

-

solubility

1 M HCl: 100 mg/mL

solubility

1 M HCl: 50 mg/mL, clear, colorless to faintly yellow

solubility

1 M HCl: 0.5 g/10 mL, clear, colorless

solubility

-

color

white to off-white

color

white to off-white

color

white

color

-


Still not finding the right product?

Explore all of our products under L-Cystine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Transport Interaction of t-Cystine and L-Glutamate in Human Diploid
Fibroblasts in Culture
Shiro Bannai and Emi Kitamur
The Journal of Biological Chemistry, 255(6), 2372- 2376 (1980)
Studies on Analogues of L-Cysteine and L-Cystine
Austin S and Leif G
Blood (2018)
Mutation of Ataxia?Telangiectasia Mutated is Associated with Dysfunctional Glutathione Homeostasis in Cerebellar Astroglia
Andrew Campbell
Glia, 64(2), 227-239 (2016)



Global Trade Item Number

SKUGTIN
C7602-25G04061833513101
C7602-1KG04061833513088
C7602-100G04061833513064
C7602-10MG04061833513071

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters