Skip to Content
Merck
All Photos(5)

Key Documents

C8755

Sigma-Aldrich

L-Cystine

≥98% (TLC)

Synonym(s):

(R,R)-3,3′-Dithiobis(2-aminopropionicacid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30
Beilstein:
1728094
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Cystine, ≥98% (TLC), crystalline

Assay

≥98% (TLC)

form

crystalline

color

white to off-white

mp

>240 °C (dec.) (lit.)

solubility

1 M HCl: 50 mg/mL, clear, colorless to faintly yellow

application(s)

cell analysis
peptide synthesis

storage temp.

room temp

SMILES string

N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

LEVWYRKDKASIDU-IMJSIDKUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Cystine has been used in in vitro cystine solubility assay to identify potential drugs that influence cystine solubility. It is also used as a supplement of phosphate-buffered saline to slice and wash periprosthetic tissues.

Biochem/physiol Actions

Cysteine is the source of disulfide linkages in proteins and has a role in sulfur transport. It undergoes rapid oxidation to form cystine at physiological pH. L-cystine is crucial for oxygen production and low density lipoprotein modification by arterial smooth muscle cells. It also has a role in the synthesis of glutathione.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The x(c)- cystine/glutamate antiporter: a potential target for therapy of cancer and other diseases
Maisie Lo
Journal of Cellular Physiology, 215(215) (2008)
Wear particles and ions from cemented and uncemented titanium-based hip prostheses?A histological and chemical analysis of retrieval material
Susann Grosse
Journal of Biomedical Materials Research, 709?717-709?717 (2015)
The role of sulfur-containing amino acids in superoxide production and modification of low density lipoprotein by arterial smooth muscle cells.
Heinecke JW
The Journal of Biological Chemistry, 262(21), 10098-10103 (1987)
Mass Spectrometry-based in vitro Assay to Identify Drugs that Influence Cystine Solubility
Neelanjan Bose
Bio-protocol, 7(14), e2417-e2417 (2017)
Eloisa Sardella et al.
Antioxidants (Basel, Switzerland), 10(4) (2021-05-01)
Plasma Treated Water Solutions (PTWS) recently emerged as a novel tool for the generation of Reactive Oxygen and Nitrogen Species (ROS and RNS) in liquids. The presence of ROS with a strong oxidative power, like hydrogen peroxide (H2O2), has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service