C6492
Ciproxifan hydrochloride
≥98% (HPLC), solid
Synonym(s):
Cyclopropyl[4-[3-(1H-imidazol-4-yl)propoxyl]phenyl]-methanone hydrochloride
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
solid
storage condition
desiccated
under inert gas
color
off-white
solubility
DMSO: 30 mg/mL
water, high purity: 30 mg/mL
storage temp.
2-8°C
SMILES string
Cl.O=C(C1CC1)c2ccc(OCCCc3c[nH]cn3)cc2
InChI
1S/C16H18N2O2.ClH/c19-16(12-3-4-12)13-5-7-15(8-6-13)20-9-1-2-14-10-17-11-18-14;/h5-8,10-12H,1-4,9H2,(H,17,18);1H
InChI key
PWAPSKMHZQQTRJ-UHFFFAOYSA-N
Biochem/physiol Actions
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service