Skip to Content
Merck
All Photos(4)

Key Documents

D3630

Sigma-Aldrich

Diphenhydramine hydrochloride

≥98% (HPLC)

Synonym(s):

2-Diphenylmethoxy-N,N-dimethylethylamine hydrochloride, N-(2-Diphenylmethoxyethyl)-N,N-dimethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

originator

Johnson & Johnson

SMILES string

Cl[H].CN(C)CCOC(c1ccccc1)c2ccccc2

InChI

1S/C17H21NO.ClH/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;/h3-12,17H,13-14H2,1-2H3;1H

InChI key

PCHPORCSPXIHLZ-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Application

Diphenhydramine hydrochloride has been used:
as a reference compound to study the long duration of action of antihistamine bilastine in vivo[1]
to detect its embryotoxicity by embryonic stem cell test (EST) using cardiomyocyte differentiation[2] and osteoblasts differentiation[3]
as an antihistamine in experimental studies[4][5]

Biochem/physiol Actions

H1 histamine receptor antagonist
Diphenhydramine hydrochloride (DPH) is an antihistaminic agent that relieves symptoms of hypersensitive reactions.[6][7] It exhibits antimuscarinic and marked sedative effects.[8] DPH is also used to prevent nausea, vomiting and vertigo of various causes.[6] In addition, it acts as a potential therapeutic for insomnia.[7]

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiation of P2RX7 as a host-directed strategy for control of mycobacterial infection
Matty MA, et al.
eLife, 8, e39123-e39123 (2019)
Alina Tokmakova et al.
iScience, 26(4), 106422-106422 (2023-04-25)
The 25 human bitter taste receptors (TAS2Rs) are expressed on taste and extra-oral cells representing an integrated chemosensory system. The archetypal TAS2R14 is activated by > 150 topographically diverse agonists, raising the question of how this uncharacteristic accommodation is achieved for these
Reevaluation of the embryonic stem cell test
Inselman AL, et al.
International journal of regulatory science, 1(1), 32-49 (2013)
Simultaneous determination of pseudoephedrine hydrochloride and diphenhydramine hydrochloride in cough syrup by gas chromatography (GC)
Raj SV, et al.
Talanta, 46(1), 221-225 (1998)
R U Agu et al.
Pharmaceutical research, 16(9), 1380-1385 (1999-09-25)
To test two in vitro human nasal epithelial cell culture systems for their ability to screen the effects of pharmaceutical compounds on ciliary beating. Human nasal epithelial cells were cultured as monolayer and in a sequential monolayer-suspension culture with in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service