Recommended Products
Assay
≥98% (TLC)
Quality Level
form
powder
solubility
water: 50 mg/mL, clear to very slightly hazy, colorless
storage temp.
−20°C
SMILES string
Cl.N[C@H](CCl)C(O)=O
InChI
1S/C3H6ClNO2.ClH/c4-1-2(5)3(6)7;/h2H,1,5H2,(H,6,7);1H/t2-;/m1./s1
InChI key
IENJPSDBNBGIEL-HSHFZTNMSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Proceedings of the National Academy of Sciences of the United States of America, 71(2), 417-421 (1974-02-01)
The D- and L-isomers of beta-chloroalanine inhibit the growth of Diplococcus pneumoniae, Streptococcus pyogenes, Bacillus subtilis, and Escherichia coli. With pneumococcus the inhibition by beta-chloro-D-alanine is completely prevented by either D-alanine or D-alanyl-D-alanine, while L-alanine is not effective in preventing
Experimental parasitology, 122(2), 106-111 (2009-03-03)
The sphingolipids biosynthesis pathway generates bioactive molecules crucial to the regulation of physiological processes. We have recently reported that DAG (diacylglycerol) generated during sphingomyelin synthesis, plays an important role in PKC (protein kinase C) activation, necessary for the transit through
Biochemical pharmacology, 65(2), 181-192 (2002-12-31)
The mitochondrial and cytosolic branched-chain aminotransferases (BCAT(m) and BCAT(c)) are homodimers in the fold type IV class of pyridoxal 5'-phosphate-containing enzymes that also contains D-amino acid aminotransferase and 4-amino-4-deoxychorismate lyase (a beta-lyase). Recombinant human BCAT(m) and BCAT(c) were shown to
The Biochemical journal, 368(Pt 1), 253-261 (2002-07-26)
Rat liver mitochondrial aspartate aminotransferase (a homodimer) was shown to catalyse a beta-lyase reaction with three nephrotoxic halogenated cysteine S-conjugates [ S -(1,1,2,2-tetrafluoroethyl)-L-cysteine, S -(1,2-dichlorovinyl)-L-cysteine and S -(2-chloro-1,1,2-trifluoroethyl)-L-cysteine], and less effectively so with a non-toxic cysteine S-conjugate [benzothiazolyl-L-cysteine]. Transamination competes
Journal of magnetic resonance (San Diego, Calif. : 1997), 165(2), 237-247 (2003-12-04)
A new approach for the isotopic labeling of proteins is proposed that aims to facilitate side chain resonance assignments. Residue-correlated (RC) labeling is achieved by the expression of a protein on a medium containing a mixture of labeled, e.g., [U-13C,15N]amino
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service