Skip to Content
Merck
All Photos(2)

Documents

Z0878

Sigma-Aldrich

Zaprinast

solid

Synonym(s):

1,4-Dihydro-5-(2-propoxyphenyl)-7H-1,2,3-triazolo(4,5-d)pyrimidin-7-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13N5O2
CAS Number:
Molecular Weight:
271.27
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.11 mg/mL
DMSO: 10 mg/mL
H2O: insoluble

SMILES string

CCCOc1ccccc1C2=NC(=O)c3nn[nH]c3N2

InChI

1S/C13H13N5O2/c1-2-7-20-9-6-4-3-5-8(9)11-14-12-10(13(19)15-11)16-18-17-12/h3-6H,2,7H2,1H3,(H2,14,15,16,17,18,19)

InChI key

REZGGXNDEMKIQB-UHFFFAOYSA-N

Gene Information

Application

Zaprinast has been used to determine its inhibitory effect on the meiosis of oocytes by germinal vesicle breakdown (GVBD) assay and to test the inhibitory effect of it on the retinal cells by immunohistochemistry.

Biochem/physiol Actions

Zaprinast insubstantially inhibits phosphodiesterase 10 and 11 (PDE10 and PDE11). It also exhibits vasodilating effects.
Selective inhibitor of cGMP-specific phosphodiesterases V and VI (PDE5/6) and an agonist at the G protein-coupled receptor 35 (GPR35).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Picconi et al.
Brain : a journal of neurology, 134(Pt 2), 375-387 (2010-12-25)
The aim of the present study was to evaluate the role of the nitric oxide/cyclic guanosine monophosphate pathway in corticostriatal long-term depression induction in a model of levodopa-induced dyskinesia in experimental parkinsonism. Moreover, we have also analysed the possibility of
Xinhua Zhang et al.
American journal of physiology. Endocrinology and metabolism, 302(2), E243-E253 (2011-10-27)
Testosterone (T) plays a permissive role in the development of benign prostatic hyperplasia (BPH), and phosphodiesterase 5 inhibitors (PDE5is) have been found to be effective for BPH and lower urinary tract symptoms (LUTS) in clinical trials. This study investigated the
Selective pulmonary vasodilation induced by aerosolized zaprinast
Ichinose F, et al.
Anesthesiology, 88(2), 410-416 (1998)
Resumption of meiosis induced by meiosis-activating sterol has a different signal transduction pathway than spontaneous resumption of meiosis in denuded mouse oocytes cultured in vitro
Faerge I, et al.
Biology of Reproduction, 65(6), 1751-1758 (2001)
A Sahaboglu et al.
Cell death & disease, 4, e488-e488 (2013-02-09)
For most neurodegenerative diseases the precise duration of an individual cell's death is unknown, which is an obstacle when counteractive measures are being considered. To address this, we used the rd1 mouse model for retinal neurodegeneration, characterized by phosphodiesterase-6 (PDE6)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service