Skip to Content
Merck
All Photos(2)

Documents

C3662

Sigma-Aldrich

Cyclosporin A

from Tolypocladium inflatum, ≥95% (HPLC), solid, Calcineurin inhibitor

Synonym(s):

Antibiotic S 7481F1, Cyclosporine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
Beilstein:
3647785
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid

biological source

Tolypocladium inflatum

Quality Level

Assay

≥95% (HPLC)

form

solid

color

white

solubility

dichloromethane: 10 mg/mL
ethanol: 10 mg/mL
DMSO: 50 mg/mL
chloroform: 6 mg/mL
H2O: insoluble

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

originator

Novartis

storage temp.

2-8°C

SMILES string

CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C

InChI

1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1

InChI key

PMATZTZNYRCHOR-CGLBZJNRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A fungal metabolite possessing potent immunosuppressive properties. It inhibits the T-cell receptor signal transduction pathway via the formation of cyclosporin A−cyclophilin complex that inhibits calcineurin (protein phosphatase 2B). Inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα. Can block cytochrome c release from mitochondria.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphoprotein Phosphatases (Serine/Threonine) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peu Santra et al.
Disease models & mechanisms, 14(7) (2021-07-24)
The vacuolar-type H+-ATPase (V-ATPase) is a multi-subunit proton pump that regulates cellular pH. V-ATPase activity modulates several cellular processes, but cell-type-specific functions remain poorly understood. Patients with mutations in specific V-ATPase subunits can develop sensorineural deafness, but the underlying mechanisms
Mi-Yeon Jang et al.
Journal of medicinal chemistry, 54(2), 655-668 (2010-12-22)
Herein we describe the synthesis and in vitro and in vivo activity of thiazolo[5,4-d]pyrimidines as a novel class of immunosuppressive agents, useful for preventing graft rejection after organ transplantation. This research resulted in the discovery of a series of compounds
Xiao-Ling Liu et al.
Bioorganic & medicinal chemistry, 16(1), 171-180 (2007-10-30)
A library of chalcones with basic functionalities were screened for inhibition of P-glycoprotein (Pgp, ABCB1) by the calcein-AM accumulation assay on MDCKII/MDR1 cells. Three members that had ring A substituted with 5-(1-ethylpiperidin-4-yl) and 2,4-dimethoxy groups were found to increase calcein-AM
Lotte Coelmont et al.
Antimicrobial agents and chemotherapy, 53(3), 967-976 (2008-12-24)
Debio 025 is a potent inhibitor of hepatitis C virus (HCV) replication (J. Paeshuyse et al., Hepatology 43:761-770, 2006). In phase I clinical studies, monotherapy (a Debio 025 dose of 1,200 mg twice a day) resulted in a mean maximal
Javier Koh et al.
Nature communications, 10(1), 2820-2820 (2019-06-30)
Bats are unusual mammals, with the ability to fly, and long lifespans. In addition, bats have a low incidence of cancer, but the mechanisms underlying this phenomenon remain elusive. Here we discovered that bat cells are more resistant than human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service