Skip to Content
Merck
All Photos(1)

Key Documents

A9666

Sigma-Aldrich

Azaserine-Hypoxanthine 50x

Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma

Synonym(s):

O-Diazoacetyl-L-serine-hypoxanthine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

biological source

synthetic (organic)

Quality Level

grade

Hybri-Max

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

shipped in

dry ice

storage temp.

−20°C

General description

Azaserine-Hypoxanthine is used to stimulate cell proliferation in cell culture.

Application

Azaserine-Hypoxanthine 50x has been used in hybridoma cell culture.
Qualified for use in hybridoma cell culture applications as a selection agent.

Reconstitution

Reconstitute contents of vial with 10 mL sterile cell culture medium. Stock solution is sufficient to prepare 500 mL medium. Final working concentration: 5.7 μM azaserine, 100 μM hypoxanthine.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Mutagenicity Test Procedures, 350-350 (2012)
Receptor editing: an approach by autoreactive B cells to escape tolerance.
Gay D,et al.
The Journal of Experimental Medicine, 177(4), 999-1008 (1993)
Handbook of Mutagenicity Test Procedures, 350-350 (2012)
Michael X Henderson et al.
Neurobiology of disease, 136, 104712-104712 (2019-12-15)
Parkinson's disease (PD) and dementia with Lewy bodies (DLB) are progressive neurodegenerative diseases for which there is no disease-modifying treatment. PD and DLB are characterized by aggregation of the synaptic protein α-synuclein, and there is compelling evidence to suggest that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service