Skip to Content
Merck
All Photos(1)

Key Documents

C-28015

Sigma-Aldrich

Mesenchymal Stem Cell Neurogenic Differentiation Medium

Ready-to-use kit including Basal Medium and SupplementMix, 100 ml

Synonym(s):

Mesenchymal Stem Cell Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

packaging

pkg of 100 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

General description

Mesenchymal Stem Cell Neurogenic Differentiation Medium (Ready-to-use)

Application

Mesenchymal Stem Cell Neurogenic Differentiation Medium is a defined and serum-free system for the directed differentiation of mesenchymal stem cells (MSC) from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue into neurogenic lineages. Recommended for:Human Mesenchymal Stem Cells from Bone Marrow (hMSC-BM)Human Mesenchymal Stem Cells from Umbilical Cord Matrix (hMSC-UC)Human Mesenchymal Stem Cells from Adipose Tissue (hMSC-AT)

Quality

All lots of PromoCell Mesenchymal Stem Cell Media are subjected to comprehensive quality control tests using primary human mesenchymal stem cells. Each lot of MSC Differentiation Media is tested for the capacity to induce directed differentiation into the respective lineages in MSC. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service