Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.71
UNSPSC Code:
12352207
form
liquid
packaging
pkg of 100 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
storage temp.
2-8°C
General description
Mesenchymal Stem Cell Chondrogenic Differentiation Medium w/o Inducers (Ready-to-use)
Application
Mesenchymal Stem Cell Chondrogenic Differentiation Medium is a defined and serum-free system for the directed differentiation of mesenchymal stem cells (MSC) from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue into chondrogenic lineages.Recommended for:Human Mesenchymal Stem Cells from Bone Marrow (hMSC-BM)Human Mesenchymal Stem Cells from Umbilical Cord Matrix (hMSC-UC)Human Mesenchymal Stem Cells from Adipose Tissue (hMSC-AT)Note: this MSC Chondrogenic Differentiation Medium w/o Inducers must be supplemented with adequate chondrogenic inducers by the customer.
Preparation Note
Click here for more information.
Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.
Analysis Note
All lots of PromoCell Mesenchymal Stem Cell Media are subjected to comprehensive quality control tests using primary human mesenchymal stem cells. Each lot of MSC Differentiation Media is tested for the capacity to induce directed differentiation into the respective lineages in MSC. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Still not finding the right product?
Explore all of our products under Mesenchymal Stem Cell Chondrogenic Differentiation Medium
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.