Skip to Content
Merck
All Photos(1)

Key Documents

C-24400

Sigma-Aldrich

Melanocyte Growth Medium M2

Basal Medium, 500 ml

Synonym(s):

Recommended with C-39420

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

form

aqueous solution

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Melanocyte M2 Basal Medium (supplements sold separately)

To form a complete medium, it should be combined with the media supplements: C-39420

Application

Melanocyte Growth Medium M2 is a serum-free and PMA (Phorbol Myristate Acetate)-free Medium that was developed for the optimized in vitro cultivation of human melanocytes. It is also suitable for human pigment epithelial cells, neuronal cells, and melanoma cells. The medium is optimized for primary human cells, but can also be used for equine, porcine, mouse, and rat melanocytes.Recommended for:Normal Human Epidermal Melanocytes (NHEM), juvenile foreskin, cultured in M2 MediumNormal Human Epidermal Melanocytes (NHEM), adult donor, cultured in M2 Medium

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium (Ready-to-use): Includes phenol red-free Basal Medium and SupplementMixSupplementMix: Contains all media supplements premixed in one vial

Quality

All lots of PromoCell Melanocyte Growth Medium M2 are subjected to comprehensive quality control tests using primary human melanocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested melanocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark immediately after arrival. Do not freeze the Basal Medium. If stored properly, the medium is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C

Subculture Routine

Click here for more information.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service