C-39420
Melanocyte Growth Medium M2
SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml
Synonym(s):
Recommended with C-24400 or C-24405
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.71
Recommended Products
form
aqueous solution
packaging
pkg of 1 vial
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Melanocyte Growth Medium M2 SupplementMix (basal medium sold separately)
To form a complete medium, it should be combined with the basal medium: C-24400 or C-24405
To form a complete medium, it should be combined with the basal medium: C-24400 or C-24405
Application
Melanocyte Growth Medium M2 is a serum-free and PMA (Phorbol Myristate Acetate)-free Medium that was developed for the optimized in vitro cultivation of human melanocytes. It is also suitable for human pigment epithelial cells, neuronal cells, and melanoma cells. The medium is optimized for primary human cells, but can also be used for equine, porcine, mouse, and rat melanocytes.Recommended for:Normal Human Epidermal Melanocytes (NHEM), juvenile foreskin, cultured in M2 MediumNormal Human Epidermal Melanocytes (NHEM), adult donor, cultured in M2 Medium
Components
Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium (Ready-to-use): Includes phenol red-free Basal Medium and SupplementMixSupplementMix: Contains all media supplements premixed in one vial
Quality
All lots of PromoCell Melanocyte Growth Medium M2 are subjected to comprehensive quality control tests using primary human melanocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested melanocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Storage and Stability
Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.
Subculture Routine
Click here for more information.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service