Skip to Content
Merck
All Photos(1)

Documents

C1224

Sigma-Aldrich

Caspase 3 human

≥90% (SDS-PAGE), recombinant, expressed in E. coli (C-terminal histidine-tagged), buffered aqueous glycerol solution, ≥1.0 units/mg protein

Synonym(s):

Apopain, CPP32, Yama

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in E. coli (C-terminal histidine-tagged)

Quality Level

Assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

≥1.0 units/mg protein

mol wt

~30 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CASP3(836)

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Apoptosis

Human recombinant C-terminal histidine tagged caspase 3 is a fully active protein consisting of 17 kDa and 13.5 kDa subunits; the 13.5 kDa subunit contains the histidine tag.

Application

Caspase 3 human has been used to digest human r-vimentin. It has also been used in caspase-3 inhibition assay and protease inhibition assay.

This product may also be used in cell-based apoptosis assay.

Biochem/physiol Actions

Caspase 3 is a member of the CED-3 subfamily of caspases and is responsible for the cleavage of many key proteins such as the nuclear enzyme poly(ADP-ribose) polymerase (PARP), the inhibitor of caspase-activated deoxyribonuclease (ICAD), and gelsolin, a protein involved in apoptosis regulation. Caspase 3 is considered to be an effector caspase, activating pro-caspase 6 and pro-caspase 9 in vitro. Caspase 3 can be activated by caspase 8, caspase 6, and granzyme B.

Unit Definition

One unit will cleave 1.0 μmole of N-acetyl-Asp-Glu-Val-Asp-pNA per min at pH 7.4 at 25 °C.

Physical form

Solution in 10% (w/v) glycerol containing 50 mM HEPES, pH 7.4, 100 mM NaCl, 10 mM DTT, 1 mM EDTA, 0.1% CHAPS

Preparation Note

Caspase 3 is synthesized as a 32 kDa proenzyme. The active enzyme is a heterodimer of two large (17 kDa) subunits and two small (12 kDa) subunits.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cross-presentation of caspase-cleaved apoptotic self antigens in HIV infection
Rawson PM, et al.
Nature Medicine, 13(12), 1431-1431 (2007)
A peptide-based positron emission tomography probe for in vivo detection of caspase activity in apoptotic cells
Hight MR, et al.
Clinical Cancer Research, 20(8), 2126-2135 (2014)
Deju Ye et al.
Nature chemistry, 6(6), 519-526 (2014-05-23)
Directed self-assembly of small molecules in living systems could enable a myriad of applications in biology and medicine, and already this has been used widely to synthesize supramolecules and nano/microstructures in solution and in living cells. However, controlling the self-assembly
Petra Procházková et al.
PloS one, 9(10), e109900-e109900 (2014-10-04)
Iron homeostasis in cells is regulated by iron regulatory proteins (IRPs) that exist in different organisms. IRPs are cytosolic proteins that bind to iron-responsive elements (IREs) of the 5'- or 3'-untranslated regions (UTR) of mRNAs that encode many proteins involved
Regulation of spermatogenic cell apoptosis by the pro-apoptotic proteins in the testicular tissues of mammalian and avian species
Zakariah M, et al.
Animal Reproduction Science (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service