Skip to Content
Merck
All Photos(1)

Documents

A1086

Sigma-Aldrich

N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin

≥97% (HPLC), powder

Synonym(s):

Ac-DEVD-AMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H37N5O13
CAS Number:
Molecular Weight:
675.64
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ~65%

color

white

solubility

DMSO: 10 mM

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(C)=O)C(=O)N[C@@H](CC(O)=O)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C30H37N5O13/c1-13(2)26(35-27(44)18(7-8-22(37)38)33-29(46)19(11-23(39)40)31-15(4)36)30(47)34-20(12-24(41)42)28(45)32-16-5-6-17-14(3)9-25(43)48-21(17)10-16/h5-6,9-10,13,18-20,26H,7-8,11-12H2,1-4H3,(H,31,36)(H,32,45)(H,33,46)(H,34,47)(H,35,44)(H,37,38)(H,39,40)(H,41,42)/t18-,19-,20-,26-/m0/s1

InChI key

ALZSTTDFHZHSCA-RNVDEAKXSA-N

Application

N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin has been used as a substrate for caspase-3 assay in human colon cancer cell line and in bone marrow mesenchymal stem cells.

Biochem/physiol Actions

Fluorogenic substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice Dreser et al.
Cell death and differentiation, 24(10), 1655-1671 (2017-06-18)
Amyotrophic lateral sclerosis (ALS) is characterized by the selective degeneration of motor neurons (MNs) and their target muscles. Misfolded proteins which often form intracellular aggregates are a pathological hallmark of ALS. Disruption of the functional interplay between protein degradation (ubiquitin
Panagiotis Kratimenos et al.
Scientific reports, 7(1), 16664-16664 (2017-12-02)
We have previously shown that cerebral Hypoxia-ischemia (HI) results in activation of Src kinase in the newborn piglet brain. We investigated the regulatory mechanism by which the pre-apoptotic proteins translocate from mitochondria to the cytosol during HI through the Src
Autophagy induced by ionizing radiation promotes cell death over survival in human colorectal cancer cells
Classen F, et al.
Experimental Cell Research, 374(1), 29-37 (2018)
Yi-Xin Wu et al.
Oncotarget, 7(47), 77622-77634 (2016-10-22)
Bortezomib (BTZ), a proteasome inhibitor, is the first proteasome inhibitor to be used in clinical practice. Here we investigated the mechanisms underlying acquired bortezomib resistance in hepatocellular carcinoma (HCC) cells. Using stepwise selection, we established two acquired bortezomib-resistant HCC cell
Saravana P Selvanathan et al.
Nucleic acids research, 47(18), 9619-9636 (2019-08-09)
Connections between epigenetic reprogramming and transcription or splicing create novel mechanistic networks that can be targeted with tailored therapies. Multiple subunits of the chromatin remodeling BAF complex, including ARID1A, play a role in oncogenesis, either as tumor suppressors or oncogenes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service