Skip to Content
Merck
All Photos(4)

Key Documents

B0385

Sigma-Aldrich

Beclomethasone

≥99%

Synonym(s):

9α-Chloro-11β,17α,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione, 9α-Chloro-16β-methyl-1,4-pregnadiene-11β,17α,21-triol-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29ClO5
CAS Number:
Molecular Weight:
408.92
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99%

form

powder

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

[H][C@]12C[C@@H](C)[C@@](O)(C(=O)CO)[C@]1(C)C[C@@H](O)[C@@]3(Cl)[C@]2([H])CCC4=CC(=O)C=C[C@@]34C

InChI

1S/C22H29ClO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12?,15?,16?,17?,19?,20?,21-,22?/m0/s1

InChI key

NBMKJKDGKREAPL-WOOYFSPWSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

Beclomethasone has been used in MS−MS (mass spectrometry) methods for the analysis of β-agonists, corticosteroids, chloramphenicol and penicillins. It has also been used to find the differential response of glucocorticoids in larval regeneration model.

Biochem/physiol Actions

Beclomethasone is an anti-inflammatory glucocorticoid.
It helps to decrease airway hyperresponsiveness. It also helps to regulate symptoms of asthma.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufei Xie et al.
Disease models & mechanisms, 12(5) (2019-05-11)
Glucocorticoid drugs are widely used to treat immune-related diseases, but their use is limited by side effects and by resistance, which especially occurs in macrophage-dominated diseases. In order to improve glucocorticoid therapies, more research is required into the mechanisms of
A comparative analysis of changes in pMDI drug dose delivery before and after detergent coating using five antistatic valved holding chambers.
Paul Hagedoorn et al.
The journal of allergy and clinical immunology. In practice, 8(3), 1124-1125 (2019-10-09)
Lydia J Finney et al.
The Journal of allergy and clinical immunology, 147(2), 510-519 (2020-10-18)
The mechanisms underlying altered susceptibility and propensity to severe Coronavirus disease 2019 (COVID-19) disease in at-risk groups such as patients with chronic obstructive pulmonary disease (COPD) are poorly understood. Inhaled corticosteroids (ICSs) are widely used in COPD, but the extent
Alternate glucocorticoid receptor ligand binding structures influence outcomes in an in vivo tissue regeneration model
Sengupta S, et al.
Comparative Biochemistry and Physiology. Toxicology & Pharmacology : CBP, 156(2), 121-129 (2012)
Sílvia M Illamola et al.
Frontiers in pharmacology, 10, 1483-1483 (2020-01-31)
About 80% of the consumers worldwide use herbal medicine (HMs) or other natural products. The percentage may vary significantly (7%-55%) among pregnant women, depending upon social status, ethnicity, and cultural traditions. This manuscript discusses the most common HMs used by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service