Skip to Content
Merck
All Photos(1)

Key Documents

B7510

Sigma-Aldrich

Beauvericin

≥97% (HPLC), suitable for HPLC

Synonym(s):

cyclo(D-α-Hydroxyisovaleryl-L-N-methyl-Phe)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H57N3O9
CAS Number:
Molecular Weight:
783.95
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Beauvericin, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder or crystals

technique(s)

HPLC: suitable

color

white to off-white

storage temp.

2-8°C

SMILES string

CC(C)C1OC(=O)C(Cc2ccccc2)N(C)C(=O)C(OC(=O)C(Cc3ccccc3)N(C)C(=O)C(OC(=O)C(Cc4ccccc4)N(C)C1=O)C(C)C)C(C)C

InChI

1S/C45H57N3O9/c1-28(2)37-40(49)46(7)35(26-32-21-15-11-16-22-32)44(53)56-39(30(5)6)42(51)48(9)36(27-33-23-17-12-18-24-33)45(54)57-38(29(3)4)41(50)47(8)34(43(52)55-37)25-31-19-13-10-14-20-31/h10-24,28-30,34-39H,25-27H2,1-9H3

InChI key

GYSCAQFHASJXRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Beauvericin is a cyclohexadepsipeptide from the fungus Beauveria bassiana that belongs to the enniatin antibiotic family. It comprises three D-hydroxyisovaleryl and N-methylphenylalanyl residues each.

Application

Beauvericin has been used as an antimicrobial bioactive compound to test its effect on engineered Saccharomyces cerevisiae strains. It has also been used as a mycotoxin standard in the reverse-phase (RP) high-pressure liquid chromatography (HPLC) to quantify mycotoxins produced by Fusarium oxysporum isolates.

Biochem/physiol Actions

Beauvericin (BEA) is a commercial entomopathogenic mycoinsecticide with antiviral, antibacterial, insecticidal, and cytotoxic properties. It elicits apoptotic and nematicidal activities as well. Beauvericin blocks multidrug efflux and Tor complex 1 (TORC1) kinase. It possesses ionophoric functionality and favors channel formation in patches of ventricular myocytes and synthetic membranes. It is shown to inhibit L-type voltage-dependent Ca2+ current in the neuroblastoma cells.
Beauvericin (BEA), a cyclohexadepsipeptide produced by the fungus Beauveria bassiana (Bals.), has antiviral, antibacterial, nematicidal, insecticidal, cytotoxic, and apoptotic activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyuk-Hwan Song et al.
International journal of food microbiology, 122(3), 296-301 (2008-02-19)
Beauvericins and enniatins are cyclohexadepsipeptide mycotoxins that exhibit phytotoxicity and insecticidal activities. In the present study, the production of beauvericin and newly found enniatins (H, I, and MK1688) was characterized in 28 Fusarium strains isolated from potato samples in Korea.
Sheng-Nan Wu et al.
Chemical research in toxicology, 15(6), 854-860 (2002-06-18)
The effects of beauverficin, a cyclodepsipeptide compound, on ion currents in a mouse neuroblastoma and rat glioma hybrid cell line, NG108-15, were investigated with the aid of the whole-cell voltage-clamp technique. Beauvericin (0.3-100 microM) reversibly produced an inhibition of L-type
K Kouri et al.
Biochimica et biophysica acta, 1609(2), 203-210 (2003-01-25)
The antibiotic Beauvericin (BEA) was previously shown to express ionophoric properties under simple experimental systems. Its channel-forming activity was examined in inside-out patches of ventricular myocytes and synthetic membranes with the patch clamp and fluorescence imaging techniques. Current transitions to
Francesca Caloni et al.
Environmental toxicology and pharmacology, 75, 103349-103349 (2020-02-07)
Beauvericin (BEA) is a natural bioactive compound, with a dual nature. On the one hand, the peculiar characteristics of its molecule confer to BEA interesting properties, such as antibacterial, antiviral, antifungal, antiparasitic, insecticidal and anticarcinogenic activities. On the other hand
Tanvi Shekhar-Guturja et al.
Antimicrobial agents and chemotherapy, 60(12), 7468-7480 (2016-11-01)
Invasive fungal infections are a leading cause of human mortality. Effective treatment is hindered by the rapid emergence of resistance to the limited number of antifungal drugs, demanding new strategies to treat life-threatening fungal infections. Here, we explore a powerful

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service