Skip to Content
Merck
All Photos(3)

Key Documents

F6513

Sigma-Aldrich

Fusaric acid

from Gibberella fujikuroi

Synonym(s):

5-Butylpicolinic acid, 5-Butylpyridine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2
CAS Number:
Molecular Weight:
179.22
Beilstein:
125804
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

biological source

Gibberella fujikuroi

Quality Level

Assay

≥98% (TLC)

form

powder

mp

95-97  °C

solubility

ethanol: 49.00-51.00 mg/mL, clear, colorless to faintly yellow

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

CCCCc1ccc(nc1)C(O)=O

InChI

1S/C10H13NO2/c1-2-3-4-8-5-6-9(10(12)13)11-7-8/h5-7H,2-4H2,1H3,(H,12,13)

InChI key

DGMPVYSXXIOGJY-UHFFFAOYSA-N

Gene Information

human ... DBH(1621)

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: pyridine

Biochem/physiol Actions

Dopamine β-hydroxylase inhibitor.

Features and Benefits

This compound is featured on the Dopamine, Norepinephrine and Epinephrine Synthesis page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dasen Xie et al.
Plant cell reports, 30(8), 1555-1569 (2011-04-21)
Hydroxyproline-rich glycoproteins (HRGPs) play a defensive role in host-pathogen interactions. However, specific roles of individual HRGPs in plant defense against pathogen are poorly understood. Changes in extracellular distribution and abundance of individual cell wall HRGPs were investigated on root sections
Robert D Stipanovic et al.
Journal of agricultural and food chemistry, 59(10), 5351-5356 (2011-04-19)
Fusarium oxysporum is a fungal pathogen that attacks many important plants. Uniquely pathogenic strains of F. oxysporum f. sp. vasinfectum were inadvertently imported into the United States on live cottonseed for dairy cattle feed. These strains produce exceptionally high concentrations
A Moretti et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 27(5), 718-728 (2010-03-31)
In a survey carried out on 87 rotted fig fruits samples collected in the Apulia region of Italy, the authors isolated 126 Fusarium strains identified as F. ramigenum (69 strains), F. solani (49), F. proliferatum (five) and three not identified.
Rouh-Mei Hu et al.
PloS one, 7(12), e51053-e51053 (2012-12-14)
Fusaric acid (5-butylpicolinic acid), a mycotoxin, is noxious to some microorganisms. Stenotrophomonas maltophilia displays an intrinsic resistance to fusaric acid. This study aims to elucidate the mechanism responsible for the intrinsic fusaric acid resistance in S. maltophilia. A putative fusaric
Leili Samadi et al.
Planta, 225(1), 223-234 (2006-07-27)
Programmed cell death (PCD), now known as apoptosis, is accompanied by specific morphological features. In this study, fusaric acid, a fusarium mycotoxin, was used to examine cell death in saffron (Crocus sativus Linnaeus) roots, using several apoptosis assays. Our results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service